Merck
  • Role of the Na(+)/K(+)-ATPase beta-subunit in peptide-mediated transdermal drug delivery.
Role of the Na(+)/K(+)-ATPase beta-subunit in peptide-mediated transdermal drug delivery.
Molecular pharmaceutics (2015-03-04)
Changli Wang, Renquan Ruan, Li Zhang, Yunjiao Zhang, Wei Zhou, Jun Lin, Weiping Ding, Longping Wen
ABSTRACT
In this work, we discovered that the Na(+)/K(+)-ATPase beta-subunit (ATP1B1) on epidermal cells plays a key role in the peptide-mediated transdermal delivery of macromolecular drugs. First, using a yeast two-hybrid assay, we screened candidate proteins that have specific affinity for the short peptide TD1 (ACSSSPSKHCG) identified in our previous work. Then, we verified the specific binding of TD1 to ATP1B1 in yeast and mammalian cells by a pull-down ELISA and an immunoprecipitation assay. Finally, we confirmed that TD1 mainly interacted with the C-terminus of ATP1B1. Our results showed that the interaction between TD1 and ATP1B1 affected not only the expression and localization of ATP1B1, but also the epidermal structure. In addition, this interaction could be antagonized by the exogenous competitor ATP1B1 or be inhibited by ouabain, which results in the decreased delivery of macromolecular drugs across the skin. The discovery of a critical role of ATP1B1 in the peptide-mediated transdermal drug delivery is of great significance for the future development of new transdermal peptide enhancers.
Sigma-Aldrich
Sodium bicarbonate-12C, 99.9 atom % 12C
USP
Levothyroxine, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
L-Thyroxine sodium salt pentahydrate, ≥98% (HPLC), powder
Sigma-Aldrich
L-Thyroxine sodium salt pentahydrate, γ-irradiated, powder, BioXtra, suitable for cell culture
Sigma-Aldrich
Bicinchoninic acid disodium salt hydrate, ≥98% (HPLC)
Sigma-Aldrich
Sodium bicarbonate, tested according to Ph. Eur.
Sigma-Aldrich
Sodium bicarbonate, powder, BioReagent, Molecular Biology, suitable for cell culture, suitable for insect cell culture
Sigma-Aldrich
Sodium bicarbonate, puriss., meets analytical specification of Ph. Eur., BP, USP, FCC, E500, 99.0-100.5%, powder
Sigma-Aldrich
Sodium bicarbonate, anhydrous, free-flowing, Redi-Dri, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Sodium bicarbonate, ACS reagent, ≥99.7%
Sigma-Aldrich
Sodium bicarbonate, anhydrous, free-flowing, Redi-Dri, ACS reagent, ≥99.7%
Sigma-Aldrich
Sodium bicarbonate, BioXtra, 99.5-100.5%
USP
Sodium bicarbonate, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Sodium bicarbonate, ReagentPlus®, ≥99.5%, powder
Sigma-Aldrich
Sodium hydrogencarbonate, −40-+140 mesh, ≥95%