Merck
  • Simultaneous quantification of three active alkaloids from a traditional Chinese medicine Ramulus Mori (Sangzhi) in rat plasma using liquid chromatography-tandem mass spectrometry.
Simultaneous quantification of three active alkaloids from a traditional Chinese medicine Ramulus Mori (Sangzhi) in rat plasma using liquid chromatography-tandem mass spectrometry.
Journal of pharmaceutical and biomedical analysis (2015-03-18)
Shuang Yang, Baolian Wang, Xuejun Xia, Xue Li, Renyun Wang, Li Sheng, Dan Li, Yuling Liu, Yan Li
ABSTRACT
Fagomine, 1-deoxynojirimycin (DNJ) and 1,4-dideoxy-1,4-imino-d-arabinitol (DAB) are the major bioactive constituents in the active fraction of alkaloids from the traditional Chinese medicine mulberry twig (Ramulus Mori, Chinese name Sang Zhi), which has a strong activity on α-glucosidase in vitro and in vivo. A rapid and sensitive liquid chromatography-tandem mass spectrometry (LC-MS/MS) method was developed and validated for the simultaneous determination of DNJ, fagomine and DAB in rat plasma. Plasma samples were prepared using a simple protein precipitation by the addition of 1% volume of Tris and two volumes of methanol-acetonitrile. The analytes and internal standard (IS, miglitol) were chromatographed in an XBridge™ amide column with a gradient mobile phase of acetonitrile-water (0.1% ammonium hydroxide) at a flow rate of 0.7mL/min. The detection was performed on a triple quadrupole tandem mass spectrometer equipped with electrospray ionization (ESI) source in positive ion mode by multiple reaction monitoring (MRM) mode. Linear detection responses were obtained for DNJ ranging from 5.00 to 5000.00ng/mL, 10.00 to 2500.00ng/mL for fagomine and DAB. The lower limits of quantification (LLOQs) were 5.00, 10.00, 10.00ng/mL for DNJ, fagomine and DAB, respectively. Intra-day and inter-day precisions (R.S.D.%) were within 10% for three analytes with accuracies (R.E.%) less than 12%. The mean recoveries of analytes were greater than 85%. All analytes were proved to be stable during the sample storage, preparation and analytic procedures. The method was successfully applied to the pharmacokinetic study of the three alkaloids in rats after oral administration of the active fraction of alkaloids from mulberry twig.
Sigma-Aldrich
Methanol, suitable for NMR (reference standard)
Sigma-Aldrich
Methylamine solution, 33 wt. % in absolute ethanol ((denatured with 1% toluene))
Sigma-Aldrich
Methylamine solution, 40 wt. % in H2O
Supelco
Methanol, analytical standard
Sigma-Aldrich
Methylamine solution, 2.0 M in THF
Sigma-Aldrich
Methylamine solution, 2.0 M in methanol
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Supelco
Acetonitrile(Neat), Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Residual Solvent - Acetonitrile(solution in DMSO), Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, ACS spectrophotometric grade, ≥99.9%
Sigma-Aldrich
Methanol, Laboratory Reagent, ≥99.6%
Supelco
Methanol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, Absolute - Acetone free
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, suitable as ACS-grade LC reagent, ≥99.9%
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%
Sigma-Aldrich
Methanol, BioReagent, ≥99.93%
Sigma-Aldrich
Methanol, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Methanol solution, suitable for NMR (reference standard), 4% in methanol-d4 (99.8 atom % D), NMR tube size 3 mm × 8 in.
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
Sigma-Aldrich
Ammonium hydroxide solution, BioUltra, ~1 M NH3 in H2O (T)
Supelco
Acetonitrile, analytical standard
Sigma-Aldrich
Acetonitrile, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Acetonitrile, HPLC Plus, ≥99.9%
Sigma-Aldrich
Acetonitrile, electronic grade, 99.999% trace metals basis
Sigma-Aldrich
Acetonitrile, suitable for DNA synthesis, ≥99.9% (GC)
Sigma-Aldrich
Acetonitrile, ACS reagent, ≥99.5%
Sigma-Aldrich
Acetonitrile, biotech. grade, ≥99.93%