Merck
  • Small expression tags enhance bacterial expression of the first three transmembrane segments of the apelin receptor.
Small expression tags enhance bacterial expression of the first three transmembrane segments of the apelin receptor.
Biochemistry and cell biology = Biochimie et biologie cellulaire (2014-06-20)
Aditya Pandey, Muzaddid Sarker, Xiang-Qin Liu, Jan K Rainey
ABSTRACT
G-protein coupled receptors (GPCRs) are inherently dynamic membrane protein modulators of various important cellular signaling cascades. The apelin receptor (AR or APJ) is a class A GPCR involved in numerous physiological processes, implicated in angiogenesis during tumour formation and as a CD4 co-receptor for entry of human immunodeficiency virus type 1 (HIV-1) to cells. Due to the lack of efficient methods to produce full-length GPCRs enriched with nuclear magnetic resonance (NMR) active (15)N, (13)C, and (or) (2)H isotopes, small GPCR fragments typically comprising 1-2 transmembrane segments are frequently studied using NMR spectroscopy. Here, we report successful overexpression of transmembrane segments 1-3 of AR (AR_TM1-3) in the C41(DE3) strain of Escherichia coli using an AT-rich gene tag previously reported to enhance cell-free expression yields. The resulting protein, with 6 additional N-terminal residues due to the expression tag, was purified using high-performance liquid chromatography (HPLC). Far UV circular dichroism spectropolarimetry demonstrates that AR_TM1-3 has the predicted ~40% α-helical character in membrane-mimetic environments. (1)H-(15)N HSQC NMR experiments imply amenability to high-resolution NMR structural characterization and stability in solution for weeks. Notably, this small expression tag approach may also be generally applicable to other membrane proteins that are difficult to express in E. coli.
Product Description
Supelco
Sodium dodecyl sulfate, dust-free pellets, suitable for electrophoresis, Molecular Biology, ≥99.0% (GC)
Supelco
DL-Dithiothreitol solution, 1 M in H2O
Sigma-Aldrich
Ammonium chloride, 99.99% trace metals basis
Sigma-Aldrich
Dimethyl sulfoxide-d6, "100%", 99.96 atom % D
Sigma-Aldrich
D-Glucose-12C6, 16O6, 99.9 atom % 16O, 99.9 atom % 12C
Sigma-Aldrich
Dimethyl sulfoxide-d6, anhydrous, 99.9 atom % D
Sigma-Aldrich
Sodium dodecyl sulfate, ACS reagent, ≥99.0%
Supelco
D-(+)-Glucose, analytical standard
USP
Sodium acetate, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Sodium azide, BioXtra
Sigma-Aldrich
Ethylenediaminetetraacetic acid disodium salt dihydrate, suitable for electrophoresis, Molecular Biology, 99.0-101.0% (titration)
Sigma-Aldrich
Sodium azide, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Ethylenediaminetetraacetic acid disodium salt dihydrate, Sigma Grade, suitable for plant cell culture, 98.5-101.5%
Sigma-Aldrich
Ethylenediaminetetraacetic acid disodium salt dihydrate, reagent grade, 98.5-101.5% (titration)
Supelco
1,1,1,3,3,3-Hexafluoro-2-propanol, derivatization grade (GC derivatization), LiChropur, ≥99.8%
Sigma-Aldrich
Sodium dodecyl sulfate, BioUltra, Molecular Biology, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ≥98.0% (GC)
Sigma-Aldrich
DL-Dithiothreitol solution, BioUltra, Molecular Biology, ~1 M in H2O
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, Molecular Biology, ≥98.5% (GC)
Sigma-Aldrich
Ammonium chloride, Molecular Biology, suitable for cell culture, ≥99.5%
Sigma-Aldrich
Sodium dodecyl sulfate, ReagentPlus®, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, 92.5-100.5% based on total alkyl sulfate content basis
Sigma-Aldrich
Ampicillin, meets USP testing specifications
Sigma-Aldrich
D-(+)-Glucose, Hybri-Max, powder, BioReagent, suitable for hybridoma
Sigma-Aldrich
D-(+)-Glucose, suitable for mouse embryo cell culture, ≥99.5% (GC)
Sigma-Aldrich
Sodium acetate, powder, BioReagent, suitable for electrophoresis, suitable for cell culture, suitable for insect cell culture, ≥99%
Sigma-Aldrich
D-(+)-Glucose, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99.5%
Sigma-Aldrich
Sodium acetate, anhydrous, Molecular Biology, ≥99%
Sigma-Aldrich
D-(+)-Glucose, ACS reagent
Sigma-Aldrich
Sodium acetate, BioXtra, ≥99.0%