Merck
  • Increased phosphorylation of ezrin is associated with the migration and invasion of fibroblast-like synoviocytes from patients with rheumatoid arthritis.
Increased phosphorylation of ezrin is associated with the migration and invasion of fibroblast-like synoviocytes from patients with rheumatoid arthritis.
Rheumatology (Oxford, England) (2014-03-07)
Youjun Xiao, Mengying Sun, Zhongping Zhan, Yujin Ye, Mingcheng Huang, Yaoyao Zou, Liuqin Liang, Xiuyan Yang, Hanshi Xu
ABSTRACT
Increasing evidence indicates that the cytoskeletal protein ezrin may play a critical role in cell motility. This study aims to investigate the role of ezrin in regulating the migration and invasion of fibroblast-like synoviocytes (FLSs) from patients with RA. Synovial tissues were obtained from 12 patients with RA and 6 with OA, and then FLSs were separated from synovial tissues. The expression of ezrin and phosphorylated ezrin (p-ezrin) was examined by Western blotting or IF staining. A specific inhibitor of ezrin phosphorylation and small interference RNA-mediated ezrin knockdown were used to inhibit the phosphorylation of ezrin. Migration and invasion of FLSs in vitro were measured by the Boyden chamber assay. Increased expression of p-ezrin protein was found in synovial tissue and FLSs in patients with RA compared with patients with OA. Stimulation with TNF-α and IL-1β increased ezrin phosphorylation in RA FLSs. Inhibition of p-ezrin protein by a specific inhibitor of phosphorylation of ezrin and small interfering RNA-mediated knockdown reduced in vitro migration and invasion, as well as actin stress fibre formation in RA FLS. Furthermore, rho kinase and p38 mitogen-activated protein kinase (MAPK) signal pathways were involved in the phosphorylation of ezrin and invasion of RA FLSs. Increased expression of p-ezrin may contribute to aberrant aggressive behaviours of RA FLSs, which are mediated by rho kinase and the p38 MAPK pathway. This suggests a novel strategy targeting phosphorylation of ezrin to prevent synovial invasiveness and joint destruction in RA.
Sigma-Aldrich
Tumor Necrosis Factor-α human, Xeno-free, recombinant, expressed in HEK 293 cells, suitable for cell culture
Sigma-Aldrich
Tumor Necrosis Factor-α human, TNF-α, recombinant, expressed in E. coli, powder, suitable for cell culture
Sigma-Aldrich
Ezrin Inhibitor, NSC668394, The Ezrin Inhibitor, NSC668394 controls the biological activity of Ezrin.
Sigma-Aldrich
TNF-α human, Animal-component free, recombinant, expressed in E. coli, suitable for cell culture
Supelco
DL-Dithiothreitol solution, 1 M in H2O
Sigma-Aldrich
DL-Dithiothreitol solution, BioUltra, Molecular Biology, ~1 M in H2O