Merck
  • Combination of precolumn nitro-reduction and ultraperformance liquid chromatography with fluorescence detection for the sensitive quantification of 1-nitronaphthalene, 2-nitrofluorene, and 1-nitropyrene in meat products.
Combination of precolumn nitro-reduction and ultraperformance liquid chromatography with fluorescence detection for the sensitive quantification of 1-nitronaphthalene, 2-nitrofluorene, and 1-nitropyrene in meat products.
Journal of agricultural and food chemistry (2015-03-13)
Kailin Deng, Tin-Yan Wong, Yinan Wang, Elvis M K Leung, Wan Chan
ABSTRACT
Carcinogenic nitropolycyclic aromatic hydrocarbons (nitro-PAHs) are ubiquitous in the ambient environment. They are emitted predominantly from internal combustion engines and by reacting polycyclic aromatic hydrocarbons with nitrogen oxide. The emerging evidence that nitro-PAHs are taken up by plants and bioaccumulatd in the food chain has aroused worldwide concerns for the potential of chronic poisoning through dietary intake. Therefore, analytical methods of high sensitivity are extremely important for assessing the risk of human exposure to nitro-PAHs. This paper describes the development of a simple and robust ultraperformance liquid chromatography coupled fluorescence detector (UPLC-FLD) method for the sensitive determination of nitro-PAHs in meat products. The method entails precolumn reduction of the otherwise nonfluorescent nitro-PAHs to amino-PAHs which strongly fluoresce for their determination by UPLC-FLD analysis. The developed method was validated for extraction efficiency, accuracy, precision, and detection limit and has been successfully applied in quantifying 1-nitronaphthalene (1-NN), 2-nitrofluorene (2-NF), and 1-nitropyrene (1-NP) in fresh and cured meat products. The results showed that the combination of Fe/H(+)-induced nitro-reduction and UPLC-FLD analysis allows sensitive quantification of 1-NN, 2-NF, and 1-NP at detection limits of 0.59, 0.51, and 0.31 μg/kg, respectively, which is at least 10 times lower than those of the existing analytical methods.
Sigma-Aldrich
1-Nitronaphthalene, 99%
1-Nitropyrene, BCR®, certified reference material
Supelco
Dichloromethane, Selectophore, ≥99.5%
Supelco
Hexane, analytical standard
1-Nitronaphthalene, BCR®, certified reference material
Sigma-Aldrich
Dichloromethane, suitable for HPLC, ≥99.9%, contains 40-150 ppm amylene as stabilizer
Sigma-Aldrich
1-Nitropyrene, ≥95%
Sigma-Aldrich
Dichloromethane, anhydrous, ≥99.8%, contains 40-150 ppm amylene as stabilizer
Sigma-Aldrich
2-Nitrofluorene, 98%
Supelco
Dichloromethane, analytical standard
Supelco
Methylene Chloride, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Dichloromethane, ACS reagent, ≥99.5%, contains 40-150 ppm amylene as stabilizer
Sigma-Aldrich
Hexane, ReagentPlus®, ≥99%
Sigma-Aldrich
Dichloromethane, ACS reagent, ≥99.5%, contains 40-150 ppm amylene as stabilizer
Sigma-Aldrich
Dichloromethane, biotech. grade, 99.9%, contains 40-150 ppm amylene as stabilizer
Sigma-Aldrich
Dichloromethane, contains 40-150 ppm amylene as stabilizer, ACS reagent, ≥99.5%
Sigma-Aldrich
Hexane, Laboratory Reagent, ≥95%
Sigma-Aldrich
Hexane, HPLC Plus, for HPLC, GC, and residue analysis, ≥95%
Sigma-Aldrich
Dichloromethane, suitable for HPLC, ≥99.8%, contains amylene as stabilizer
Sigma-Aldrich
Hexane, suitable for HPLC, ≥95%
Sigma-Aldrich
Dichloromethane, HPLC Plus, for HPLC, GC, and residue analysis, ≥99.9%, contains 50-150 ppm amylene as stabilizer