Merck
  • Neuropeptide FF attenuates RANKL-induced differentiation of macrophage-like cells into osteoclast-like cells.
Neuropeptide FF attenuates RANKL-induced differentiation of macrophage-like cells into osteoclast-like cells.
Archives of oral biology (2014-12-03)
Yulong Sun, Xiaohu Chen, Zhihao Chen, Xiaoli Ma, Dijie Li, Peng Shang, Airong Qian
ABSTRACT
Neuropeptide FF (NPFF) has been implicated in many physiological processes but not osteoclastogenesis. We previously demonstrated that NPFF modulates the viability and nitric oxide (NO) production of RAW264.7 macrophages. This study was designed to investigate the effect of NPFF on receptor activator of nuclear factor κB ligand (RANKL)-mediated differentiation of RAW264.7 cells into osteoclast-like cells. RAW264.7 cells were cultured in 96-stripwell plates or in Corning Osteo Assay Surface 96-well plates in the presence of various concentrations of NPFF with or without RANKL for 3 or 6 d. The differentiation of osteoclast-like cells was analyzed by tartrate-resistant acid phosphatase (TRAP) stain, TRAP activity and bone resorption capacity, respectively. The mRNA expression of NPFF2 receptor (NPFFR2) and osteoclast genes was evaluated by using real-time quantitative PCR which includes TRAP, RANK (receptor activator of NF-κB), Cathepsin K, MMP-9 (matrix metallopeptidase 9), Intβ3 (integrin β3) and NFATc1 (nuclear factor of activated T cells cytoplasmic 1). In addition, the influence of NPFF on the cell viability and NO release of RAW264.7 cells was measured by MTT assay and Griess method, respectively. NPFF dose-dependently inhibited RANKL-induced osteoclast-like cells differentiation including TRAP-positive cell formation, TRAP activity and bone resorption capacity. Moreover, NO release and osteoclast gene expression of osteoclast-like cells were downregulated by NPFF. In addition, NPFFR2 gene expression in osteoclast-like cells was augmented in response to NPFF treatment. Our findings showed that NPFF could attenuate osteoclast-like cells differentiation in an in vitro osteoclastogenesis model.
Sigma-Aldrich
Thiazolyl Blue Tetrazolium Bromide, 98%
Sigma-Aldrich
Thiazolyl Blue Tetrazolium Bromide, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥97.5% (HPLC)
Sigma-Aldrich
4-Nitrophenyl phosphate disodium salt hexahydrate, tablet
Sigma-Aldrich
4-Nitrophenyl phosphate disodium salt hexahydrate, tablet
Sigma-Aldrich
4-Nitrophenyl phosphate disodium salt hexahydrate, tablet
Sigma-Aldrich
HEPES, BioXtra, pH 5.0-6.5 (1 M in H2O), ≥99.5% (titration)
Sigma-Aldrich
HEPES, BioXtra, suitable for mouse embryo cell culture, ≥99.5% (titration)
Sigma-Aldrich
HEPES, ≥99.5% (titration)
Sigma-Aldrich
HEPES, BioPerformance Certified, ≥99.5% (titration), suitable for cell culture
Sigma-Aldrich
4-Nitrophenol, spectrophotometric grade
Sigma-Aldrich
Ser-Phe-Leu-Leu-Arg-Asn-Pro-Asn-Asp-Lys-Tyr-Glu-Pro-Phe, ≥97% (HPLC)
Sigma-Aldrich
4-Nitrophenyl phosphate disodium salt hexahydrate, powder, BioReagent, suitable for cell culture, ≥97%
Sigma-Aldrich
HEPES, BioUltra, Molecular Biology, ≥99.5% (T)
Sigma-Aldrich
4-Nitrophenol, ReagentPlus®, ≥99%
Sigma-Aldrich
Phosphatase substrate, 100 mg capsules
Sigma-Aldrich
Phosphatase substrate, powder
Sigma-Aldrich
4-Nitrophenol solution, 10 mM
Sigma-Aldrich
Phosphatase substrate, 5 mg tablets
Sigma-Aldrich
Phosphatase substrate, 40 mg tablets
Sigma-Aldrich
Phosphatase substrate, 40 mg capsules
Sigma-Aldrich
4-Nitrophenyl phosphate disodium salt hexahydrate, suitable for enzyme immunoassay, ≥99.0% (enzymatic)
Sigma-Aldrich
Phosphatase substrate, Suitable for manufacturing of diagnostic kits and reagents
Sigma-Aldrich
HEPES, anhydrous, free-flowing, Redi-Dri, ≥99.5%
Supelco
4-Nitrophenol, PESTANAL®, analytical standard
Supelco
HEPES, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Acetaminophen Related Compound F, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
HEPES buffer solution, 1 M in H2O
Sigma-Aldrich
L-Glutamine
Sigma-Aldrich
L-Glutamine, meets USP testing specifications, suitable for cell culture, 99.0-101.0%, from non-animal source
Sigma-Aldrich
L-Glutamine, ReagentPlus®, ≥99% (HPLC)