Merck
  • Novel in situ gelling ocular films for the opioid growth factor-receptor antagonist-naltrexone hydrochloride: fabrication, mechanical properties, mucoadhesion, tolerability and stability studies.
Novel in situ gelling ocular films for the opioid growth factor-receptor antagonist-naltrexone hydrochloride: fabrication, mechanical properties, mucoadhesion, tolerability and stability studies.
International journal of pharmaceutics (2014-12-03)
Hamdy Abdelkader, Barbara Pierscionek, Raid G Alany
ABSTRACT
Naltrexone hydrochloride (NTX) is an innovative drug used in ophthalmology for treatment of ocular surface diseases such as impaired corneal wound healing and severe dry eye. Poor chemical stability has been a major limitation for development of NTX in solution form. The aim of this study was to develop and characterise NTX in situ ocular films for enhanced chemical stability and improved ocular tolerability. The films were prepared from different amorphous polymers and characterised for physicochemical compatibility, moisture-sorption, surface pH, mechanical properties, sterilisability, surface morphology, mucoadhesion, in vitro release, conjunctival irritation and accelerated stability at 40°C/75% relative humidity for 3 months. Glycerin (GLY)-plasticised films exhibited significantly better mechanical properties, compared with polyethylene glycol (PEG) 400 and triethylcitrate (TEC)-plasticised formulations. Superior mucoadhesion was recorded for F7 and F9 plasticised with GLY and PEG 400, respectively. The stability of NTX was significantly enhanced more than 18-times, compared with the solution form. Combination of carboxymethylcellulose sodium (CMC) and sodium alginate (ALG) in a film formulation demonstrated minimal % moisture sorption, good mechanical properties, in vitro release, excellent chemical stability and minimal conjunctival irritation lending them as promising ocular formulations.
Product Description
Supelco
L-Lysine monohydrochloride, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Supelco
L-Lysine hydrochloride solution, 100 mM amino acid in 0.1 M HCl, analytical standard
Sigma-Aldrich
L-Lysine monohydrochloride, BioUltra, ≥99.5% (AT)
Sigma-Aldrich
L-Lysine monohydrochloride, reagent grade, ≥98% (HPLC)
Sigma-Aldrich
Glycerol, BioXtra, ≥99% (GC)
Sigma-Aldrich
Glycerol, tested according to Ph. Eur., anhydrous
Sigma-Aldrich
Glycerol, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for electrophoresis, ≥99% (GC)
Sigma-Aldrich
L-Lysine monohydrochloride, from non-animal source, meets EP, JP, USP testing specifications, suitable for cell culture, 98.5-101.0%
Supelco
L-Lysine monohydrochloride, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Glycerol, analytical standard
Sigma-Aldrich
Glycerol, ACS reagent, ≥99.5%
Sigma-Aldrich
Glycerol, ReagentPlus®, ≥99.0% (GC)
Sigma-Aldrich
Glycerol, Molecular Biology, ≥99.0%
Sigma-Aldrich
Glycerol, BioUltra, Molecular Biology, anhydrous, ≥99.5% (GC)
Sigma-Aldrich
Glycerol, FCC, FG
Sigma-Aldrich
Glycerin, meets USP testing specifications
Sigma-Aldrich
Glycerol, ≥99.5%
Sigma-Aldrich
Naltrexone hydrochloride
Supelco
Glycerin, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Triethyl citrate, ≥98.0% (GC)
Supelco
Triethyl citrate, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Triethyl citrate, ≥99%, FCC, FG
Sigma-Aldrich
Glycerol solution, 83.5-89.5% (T)
USP
Glycerin, United States Pharmacopeia (USP) Reference Standard
Lysine hydrochloride, European Pharmacopoeia (EP) Reference Standard
Naltrexone hydrochloride, European Pharmacopoeia (EP) Reference Standard