Merck
  • Reduction of molecular leaching from a gel matrix for the precisely controlled release of encapsulated molecules by light stimulus.
Reduction of molecular leaching from a gel matrix for the precisely controlled release of encapsulated molecules by light stimulus.
Chemical & pharmaceutical bulletin (2014-07-06)
Takaki Amamoto, Tomofumi Santa, Masaru Kato
ABSTRACT
We have developed a general method for controlling molecular functions using a photodegradable hydrogel; gels containing molecules made from such materials are capable of release and activation by light stimulus. As the elimination of molecular leaching from the gel before irradiation was a barrier to the precise control of molecular function, we optimized the monomer used in gel preparation during this study. The addition of N,N'-methylenebis(acrylamide) (MBAA) inhibited molecular leaching from the gel; the MBAA concentration is a critical factor in controlling the leaching of encapsulated molecules. We succeeded in preparing a gel that halved the leaching of small encapsulated molecules, while the leaching of large molecules, such as albumin (66 kDa) and ferritin (450 kDa), was at negligible levels, or disappeared. The on/off ratios (released amount/leached amount) of albumin and ferritin were 8 and 17, respectively.
Sigma-Aldrich
N,N,N′,N′-Tetramethylethylenediamine, ≥99.5%, purified by redistillation
Sigma-Aldrich
N,N,N′,N′-Tetramethylethylenediamine, ReagentPlus®, 99%
Sigma-Aldrich
Methanol solution, suitable for NMR (reference standard), 4% in methanol-d4 (99.8 atom % D), NMR tube size 3 mm × 8 in.
Supelco
Dichloromethane, Selectophore, ≥99.5%
Supelco
Acrylamide solution, 40% in H2O
Sigma-Aldrich
Dichloromethane, anhydrous, ≥99.8%, contains 40-150 ppm amylene as stabilizer
Sigma-Aldrich
4-(4,6-Dimethoxy-1,3,5-triazin-2-yl)-4-methylmorpholinium chloride, 95% (HPLC)
Sigma-Aldrich
Ammonium persulfate, BioUltra, ≥98.0% (RT)
Sigma-Aldrich
Fluorescein 5(6)-isothiocyanate, BioReagent, suitable for fluorescence, mixture of 2 components, ≥90% (HPLC)
Sigma-Aldrich
Dichloromethane, suitable for HPLC, ≥99.9%, contains 40-150 ppm amylene as stabilizer
Sigma-Aldrich
Sigma 7-9®, ≥99% (titration), powder or crystals
Sigma-Aldrich
N,N′-Methylenebisacrylamide solution, suitable for electrophoresis, 2% in H2O
Sigma-Aldrich
Acrylamide solution, 40%, suitable for electrophoresis, sterile-filtered
Sigma-Aldrich
N,N,N′,N′-Tetramethylethylenediamine, BioReagent, Molecular Biology, ≥99% (GC)
Sigma-Aldrich
N,N,N′,N′-Tetramethylethylenediamine, BioReagent, suitable for electrophoresis, ≥99.0%
Sigma-Aldrich
Fluorescein 5(6)-isothiocyanate, ≥90% (HPLC)
Sigma-Aldrich
Ammonium persulfate, BioXtra, ≥98.0%
Supelco
Methylene Chloride, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Ammonium persulfate, Molecular Biology, suitable for electrophoresis, ≥98%
Supelco
Dichloromethane, analytical standard
Sigma-Aldrich
Ammonium persulfate, free-flowing, Redi-Dri, reagent grade, 98%
Sigma-Aldrich
Dichloromethane, suitable for HPLC, ≥99.8%, contains amylene as stabilizer
Sigma-Aldrich
Dichloromethane, HPLC Plus, for HPLC, GC, and residue analysis, ≥99.9%, contains 50-150 ppm amylene as stabilizer
SAFC
Tromethamine
Supelco
Tromethamine, pharmaceutical secondary standard, certified reference material
Sigma-Aldrich
Dichloromethane, ACS reagent, ≥99.5%, contains 40-150 ppm amylene as stabilizer
Sigma-Aldrich
Ammonium persulfate, ACS reagent, ≥98.0%
Sigma-Aldrich
Dichloromethane, ACS reagent, ≥99.5%, contains 40-150 ppm amylene as stabilizer
Sigma-Aldrich
Ammonium persulfate, reagent grade, 98%
Sigma-Aldrich
Dichloromethane, contains 40-150 ppm amylene as stabilizer, ACS reagent, ≥99.5%