Merck
  • Increased prevalence of cardiac autonomic dysfunction at different degrees of glucose intolerance in the general population: the KORA S4 survey.
Increased prevalence of cardiac autonomic dysfunction at different degrees of glucose intolerance in the general population: the KORA S4 survey.
Diabetologia (2015-03-01)
Dan Ziegler, Andreas Voss, Wolfgang Rathmann, Alexander Strom, Siegfried Perz, Michael Roden, Annette Peters, Christa Meisinger
ABSTRACT
Cardiac autonomic nervous dysfunction (CAND) raises the risk of mortality, but the glycaemic threshold at which it develops is unclear. We aimed to determine the prevalence of, risk factors for and impact of CAND in glucose intolerance and diabetes. Among 1,332 eligible participants aged 55-74 years in the population-based cross-sectional KORA S4 study, 130 had known diabetes mellitus (k-DM), and the remaining 1,202 underwent an OGTT. Heart rate variability (HRV) and QT variability were computed from supine 5 min ECGs. In all, 565 individuals had normal glucose tolerance (NGT), 336 had isolated impaired fasting glucose (i-IFG), 72 had isolated impaired glucose tolerance (i-IGT), 151 had combined IFG-IGT (IFG-IGT) and 78 had newly detected diabetes mellitus (n-DM). Adjusted normal HRV limits were defined in the NGT population (5th and 95th percentiles). Three HRV measures were more frequently abnormal in those with k-DM, n-DM, IFG-IGT and i-IFG than in those with NGT (p < 0.05). The rates of CAND (≥2 of 4 HRV indices abnormal) were: NGT, 4.5%; i-IFG, 8.1%; i-IGT, 5.9%; IFG-IGT, 11.4%; n-DM, 11.7%; and k-DM, 17.5% (p < 0.05 vs NGT, except for i-IGT). Reduced HRV was associated with cardiovascular risk factors used to construct a simple screening score for CAND. Mortality was higher in participants with reduced HRV (p < 0.05 vs normal HRV). In the general population aged 55-74 years, the prevalence of CAND is increased not only in individuals with diabetes, but also in those with IFG-IGT and, to a lesser degree, in those with i-IFG. It is associated with mortality and modifiable cardiovascular risk factors which may be used to screen for diminished HRV in clinical practice.
Product Description
Supelco
Cholesterol, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Cholesterol solution, certified reference material, 10 mg/mL in chloroform
Sigma-Aldrich
SyntheChol® NS0 Supplement, 500 ×, synthetic cholesterol, animal component-free, aqueous solution, sterile-filtered, suitable for cell culture
Sigma-Aldrich
Cholesterol, powder, BioReagent, suitable for cell culture, ≥99%
Sigma-Aldrich
Cholesterol, Sigma Grade, ≥99%
Sigma-Aldrich
Cholesterol, from sheep wool, ≥92.5% (GC), powder
Sigma-Aldrich
Cholesterol, from lanolin, ≥99.0% (GC)
Sigma-Aldrich
Cholesterol, tested according to Ph. Eur.
SAFC
Cholesterol, from sheep wool, Controlled origin, meets USP/NF testing specifications