Merck
  • Oxidative damage markers are significantly associated with the carotid artery intima-media thickness after controlling for conventional risk factors of atherosclerosis in men.
Oxidative damage markers are significantly associated with the carotid artery intima-media thickness after controlling for conventional risk factors of atherosclerosis in men.
PloS one (2015-03-26)
Jin-Ha Yoon, Jang-Young Kim, Jong-Ku Park, Sang-Baek Ko
ABSTRACT
This study aimed to assess the association between oxidative damage markers and carotid artery intima-media thickness (CIMT) after controlling for conventional risk factors of atherosclerosis in multiple logistic regression models. Fifty-one case male participants (CIMT ≥ 0.9 mm) were enrolled during their visits to Korean Genomic Rural Cohort Study of Wonju centers between May 1 and August 31, 2011, along with 51 control participants (CIMT < 0.9 mm) selected using frequency matching by age group. The levels of oxidative damage markers, 8-hydroxy-2'-deoxyquuanosine (8-OHdG), malondialdehyde (MDA), and 8-iso-prostaglandin F2α (Isoprostane), were measured. Conditional logistic regression models were used to evaluate relative relationships between the oxidative damage markers and the risk of high CIMT. The markers of oxidative lipid (Isoprostane and MDA) and DNA (8-OHdG) damage were associated with CIMT after controlling for the conventional risk factors, including age, low density lipoprotein, body mass index, smoking history, alcohol consumption, and metabolic syndrome (ORs [95% CI] for Isoprostane: 3rd tertile, 8.47 [2.59-27.67]; for MDA: 3rd tertile, 8.47 [2.59-27.67]; for 8-OHdG: 3rd tertile, 5.58 [1.79-17.33]). When all the oxidative damage markers were incorporated in the same logistic regression model, only Isoprostane was significantly related to CIMT (OR [95% CI]: 4.22 [1.31-13.53] in 2nd tertile and 14.21 [3.34-60.56] in 3rd tertile). In this nested case-control study, the oxidative damage markers of lipid and DNA were associated with CIMT even after controlling for the conventional risk factors of cardiovascular diseases.
Sigma-Aldrich
SyntheChol® NS0 Supplement, 500 ×, synthetic cholesterol, animal component-free, sterile-filtered, aqueous solution, suitable for cell culture
Supelco
Cholesterol solution, certified reference material, 10 mg/mL in chloroform
Supelco
4,4′-Diaminodiphenylmethane, analytical standard
Sigma-Aldrich
Cholesterol, Sigma Grade, ≥99%
Sigma-Aldrich
Cholesterol, from lanolin, ≥99.0% (GC)
Sigma-Aldrich
4,4′-Diaminodiphenylmethane, ≥97.0% (GC)
Sigma-Aldrich
Cholesterol, tested according to Ph. Eur.
Sigma-Aldrich
Cholesterol, powder, BioReagent, suitable for cell culture, ≥99%
Sigma-Aldrich
Cholesterol, from sheep wool, ≥92.5% (GC), powder
Supelco
Cholesterol, Pharmaceutical Secondary Standard; Certified Reference Material
SAFC
Cholesterol, from sheep wool, Controlled origin, meets USP/NF testing specifications