Merck
  • Development and validation of a liquid-chromatography tandem mass spectrometry method to determine in vitro and in vivo histamine release.
Development and validation of a liquid-chromatography tandem mass spectrometry method to determine in vitro and in vivo histamine release.
Journal of pharmaceutical and biomedical analysis (2014-12-03)
Krishna C Chimalakonda, Eric Pang, James L Weaver, Kristina E Howard, Vikram Patel, Michael T Boyne
ABSTRACT
Histamine is an important biogenic amine involved in regulating numerous physiological and pathophysiological processes in humans and animals. To date, there have been very few studies focused on developing and validating sensitive liquid-chromatography-tandem mass spectrometric (LC-MS/MS) assays capable of quantitative trace level histamine analysis in biological matrices. In the present study, a rapid and sensitive LC-MS/MS assay, amenable to high throughput analysis was developed and validated to characterize in vitro and in vivo histamine release. The LC-MS/MS procedure incorporating deuterium labeled internal standards provides rapid resolution of histamine with excellent sensitivity, precision, and accuracy. Histamine eluted at 1.5 min and was well separated from endogenous plasma peaks. The total run time of the assay was 8.0 min. A linear (r(2) ≥ 0.99) instrument response over the entire concentration range of 1.0-1000 ng/mL was observed. Excellent accuracy (error ± 3.4%) and precision (CV ± 10%) of the assay was demonstrated, with the lower limit of quantitation (LLOQ) at 15.6 ng/mL. The validated LC-MS/MS assay was applied to determine histamine release in both in vitro and in vivo models. Peritoneal mast cells treated with prototypical degranulating agents (Compound 48/80 and Teicoplanin) showed that the two chemicals caused approximately 40% histamine release. In rats, using this assay, basal histamine plasma levels were typically under 100 ng/mL. Treatment with an agent suspected of causing anaphylactic type reactions resulted in plasma histamine levels to increase above 3000 ng/mL. The LC-MS/MS assay presented in this study can be applied to further characterize the physiological and pathophysiological role of histamine release in complex in vitro and in vivo models. Importantly, the LC-MS/MS assay may be useful in assessing active pharmaceutical ingredient-mediated degranulation and anaphylaxis as part of either a pre-market or a post-market assessment of drug products.
Sigma-Aldrich
Acetonitrile, electronic grade, 99.999% trace metals basis
Sigma-Aldrich
Formic acid, ACS reagent, ≥96%
Sigma-Aldrich
Acetonitrile, ReagentPlus®, 99%
Sigma-Aldrich
Acetonitrile, ACS reagent, ≥99.5%
Sigma-Aldrich
Formic acid, reagent grade, ≥95%
Sigma-Aldrich
Formic acid, ACS reagent, ≥88%
Sigma-Aldrich
Acetonitrile, biotech. grade, ≥99.93%
Sigma-Aldrich
Acetonitrile, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Acetonitrile, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Acetonitrile, HPLC Plus, ≥99.9%
USP
Residual Solvent Class 2 - Acetonitrile, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Deuterium hydride, extent of labeling: 96 mol% DH, 98 atom % D
Sigma-Aldrich
Formic acid, ≥95%, FCC, FG
Sigma-Aldrich
Deuterium, 99.9 atom % D
Sigma-Aldrich
Ethylenediaminetetraacetic acid, 99.995% trace metals basis
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ≥98.0% (KT)
Sigma-Aldrich
Ethylenediaminetetraacetic acid solution, 0.02% in DPBS (0.5 mM), sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ACS reagent, 99.4-100.6%, powder
Sigma-Aldrich
Ethylenediaminetetraacetic acid, BioUltra, anhydrous, ≥99% (titration)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, anhydrous, crystalline, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid, purified grade, ≥98.5%, powder
Sigma-Aldrich
Deuterium, 99.8 atom % D
Sigma-Aldrich
Ethylenediaminetetraacetic acid disodium salt solution, BioUltra, pH 8.0, ~0.5 M in H2O
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
Sigma-Aldrich
Formic acid solution, BioUltra, 1.0 M in H2O
Sigma-Aldrich
Acetonitrile, suitable for DNA synthesis, ≥99.9% (GC)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, BioUltra, ≥99.0% (KT)
Supelco
Acetonitrile, analytical standard
Sigma-Aldrich
Histamine, ≥97.0%
Supelco
Histamine, analytical standard