Merck
  • Protective effects of polyphenols-enriched extract from Huangshan Maofeng green tea against CCl4-induced liver injury in mice.
Protective effects of polyphenols-enriched extract from Huangshan Maofeng green tea against CCl4-induced liver injury in mice.
Chemico-biological interactions (2014-06-29)
Yanmang Cui, Xingbin Yang, Xinshan Lu, Jinwen Chen, Yan Zhao
ABSTRACT
The study was to characterize the polyphenolic composition, antioxidant properties, and hepatoprotective effects of a polyphenols-enriched extract (HMTP) from Huangshan Maofeng green tea. HPLC analysis showed that three predominantly polyphenolic compounds present in HMTP were epigallocatechin (271.2 μg/mg extract), rutin (239.3 μg/mg) and epicatechin (89.3 μg/mg). HMTP was shown to exhibit strong scavenging activities against DPPH, O2(-), and OH, and ferric-reducing antioxidant power in vitro. Administration of HMTP at 200, 400 and 800 mg/kg bw in mice prior to CCl4 injury significantly decreased the CCl4-induced elevation of serum ALT, AST and ALP activities, and prevented an increase in hepatic MDA levels (p<0.05). Mice with HMTP pretreatment displayed a better profile of hepatosomatic index and the improved GSH-Px and SOD activities in the liver, relative to CCl4-intoxicated mice. Liver pathological observation also confirmed the protection on CCl4-caused histological alteration, suggesting that HMTP has potential to be explored as valuable hepatoprotective function food.
Product Description
Supelco
(−)-Epicatechin, analytical standard
Sigma-Aldrich
Sodium trichloroacetate, 97%
Supelco
4,4′-Diaminodiphenylmethane, analytical standard
Hyperoside, primary reference standard
Chlorogenic acid, primary reference standard
USP
Butylated hydroxytoluene, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
4,4′-Diaminodiphenylmethane, ≥97.0% (GC)
Sigma-Aldrich
Trichloroacetic acid, BioUltra, ≥99.5% (T)
Sigma-Aldrich
Trichloroacetic acid, ACS reagent, for the determination of Fe in blood according to Heilmeyer, ≥99.5%
Sigma-Aldrich
Butylated hydroxytoluene, ≥99%, FCC, FG
Sigma-Aldrich
(−)-Epicatechin, ≥90% (HPLC)
Sigma-Aldrich
Chlorogenic acid, ≥95% (titration)
Sigma-Aldrich
(−)-Epicatechin gallate, ≥98% (HPLC), from green tea
Sigma-Aldrich
Trichloroacetic acid, ≥99.0% (titration)
Sigma-Aldrich
Trichloroacetic acid, ACS reagent, ≥99.0%
Sigma-Aldrich
Nitrotetrazolium Blue chloride, powder, electrophoresis grade
Sigma-Aldrich
Nitrotetrazolium Blue chloride, ≥90.0% (HPLC)
Sigma-Aldrich
Trichloroacetic acid, BioXtra, ≥99.0%
Sigma-Aldrich
(−)-Epicatechin, ≥98% (HPLC), from green tea
Sigma-Aldrich
Trichloroacetic acid, suitable for electrophoresis, suitable for fixing solution (for IEF and PAGE gels), ≥99%
Supelco
Butylated Hydroxytoluene, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Edelfosine, ≥95% (HPLC)
Epicatechin gallate, primary reference standard
Epicatechin, primary reference standard
Sigma-Aldrich
Quercetin, ≥95% (HPLC), solid
Supelco
Glutathione, Pharmaceutical Secondary Standard; Certified Reference Material
Glutathione, European Pharmacopoeia (EP) Reference Standard
Caffeine for system suitability, European Pharmacopoeia (EP) Reference Standard
Supelco
Quercetin, Pharmaceutical Secondary Standard; Certified Reference Material
USP
Quercetin, United States Pharmacopeia (USP) Reference Standard