Merck
  • Biosynthesis of the new broad-spectrum lipopeptide antibiotic paenibacterin in Paenibacillus thiaminolyticus OSY-SE.
Biosynthesis of the new broad-spectrum lipopeptide antibiotic paenibacterin in Paenibacillus thiaminolyticus OSY-SE.
Research in microbiology (2014-03-13)
En Huang, Yaoqi Guo, Ahmed E Yousef
ABSTRACT
Paenibacterin is a novel lipopeptide antibiotic with potent activity against Gram-negative and Gram-positive human pathogens. The antibiotic consists of a cyclic 13-residue peptide and an N-terminal C₁₅ fatty acyl chain. To elucidate the biosynthesis of paenibacterin, we determined the whole genome sequence of the producer strain Paenibacillus thiaminolyticus OSY-SE, and the function of the peptide synthetase was confirmed experimentally. The gene cluster of paenibacterin was identified within a 52-kb DNA region, encoding thee non-ribosomal peptide synthetases, PbtA, PbtB and PbtC, and two ABC-transporters, PbtD and PbtE. Both PbtA and PbtB consist of five modules, whereas PbtC comprises three modules. Each of these 13 modules consists of three essential domains (condensation-adenylation-thiolation) and assembles an amino acid into the paenibacterin peptide. Selected adenylation domains in the NRPS were cloned and expressed in Escherichia coli; the substrate specificity of each recombinant A-domain was studied in vitro by protein function analysis. The presence of four epimerization domains in paenibacterin peptide synthetases suggests that Orn₁, Orn₄, Lys₇ and Ser₈ in the paenibacterin molecule have d-configuration; the absolute configuration of two ornithine residues in paenibacterin was confirmed by chiral amino acid analysis using Marfey's reagents. Taken together, the findings enabled us to propose the biosynthetic pathway of paenibacterin.
Product Description
USP
Sodium bicarbonate, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Sodium bicarbonate, ReagentPlus®, ≥99.5%, powder
Sigma-Aldrich
Sodium bicarbonate, ACS reagent, ≥99.7%
Sigma-Aldrich
Sodium bicarbonate, anhydrous, free-flowing, Redi-Dri, ACS reagent, ≥99.7%
Sigma-Aldrich
Sodium bicarbonate, puriss., meets analytical specification of Ph. Eur., BP, USP, FCC, E500, 99.0-100.5%, powder
Sigma-Aldrich
Sodium bicarbonate, anhydrous, free-flowing, Redi-Dri, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Sodium hydrogencarbonate, −40-+140 mesh, ≥95%
Sigma-Aldrich
Sodium bicarbonate, BioXtra, 99.5-100.5%
Sigma-Aldrich
Sodium bicarbonate, powder, BioReagent, Molecular Biology, suitable for cell culture, suitable for insect cell culture
Sigma-Aldrich
Nα-(2,4-Dinitro-5-fluorophenyl)-L-alaninamide, powder
Supelco
Nα-(2,4-Dinitro-5-fluorophenyl)-L-alaninamide, derivatization grade (chiral), LiChropur, ≥99.0%
Sigma-Aldrich
Sodium bicarbonate, tested according to Ph. Eur.
Supelco
Malachite Green chloride, analytical standard
Sigma-Aldrich
DL-Tyrosine, 99%
Sigma-Aldrich
Nitrogen, ≥99.998%
Sigma-Aldrich
Sodium bicarbonate-12C, 99.9 atom % 12C
Tyrosine, European Pharmacopoeia (EP) Reference Standard