Merck
  • Physicochemical characterisation of four cherry species (Prunus spp.) grown in China.
Physicochemical characterisation of four cherry species (Prunus spp.) grown in China.
Food chemistry (2014-12-04)
Jinping Cao, Qing Jiang, Juanying Lin, Xian Li, Chongde Sun, Kunsong Chen
ABSTRACT
The physicochemical characteristics of four cherry species (Prunus avium, Prunus cerasus, Prunus pseudocerasus and Prunus tomentosa) were evaluated. Inter-species variability was greater than intra-species differences. Glucose and fructose were the main sugars, and malic acid was the main organic acid in all species. Combining HPLC-DAD and LC-ESI-MS/MS technologies, total 25 phenolic components were preliminarily identified. P. avium was characterised by high fruit weight, edible proportion, sugar content and low acid content, which made it suitable for fresh eating. P. cerasus was high in acid content and anthocyanins content, making it a good processing species. P. pseudocerasus had rich flavonols varieties and high proportion of hydrocinnamic acids. P. tomentosa was characterised by high total phenolics content (especially flavonols and tannins) and antioxidant activity, indicating a great developmental potential as a health fruit. The results of the present study might provide theoretical guidance for the further development and utilisation of cherries.
Sigma-Aldrich
CPNQ, ≥98% (HPLC), solid
Supelco
(−)-Epicatechin, analytical standard
USP
Sodium acetate, United States Pharmacopeia (USP) Reference Standard
Epicatechin, primary reference standard
Sigma-Aldrich
Potassium chloride, tested according to Ph. Eur.
Supelco
Potassium chloride solution, conductance standard A acc. to ISO 7888, 0.1 M KCl
Supelco
Potassium chloride solution, conductance standard C acc. to ISO 7888, 0.001 M KCl
Supelco
ISA (ionic strength adjustment solution: 1 M KCl), 1 M KCl
Sigma-Aldrich
Sodium acetate solution, BioUltra, Molecular Biology, ~3 M in H2O
Supelco
Potassium chloride solution, conductance standard B acc. to ISO 7888, 0.01 M KCl
Sigma-Aldrich
Potassium chloride, BioUltra, Molecular Biology, ≥99.5% (AT)
Sigma-Aldrich
Sodium Acetate Anhydrous, >99%, FG
Sigma-Aldrich
Potassium chloride, ≥99.99% trace metals basis
Sigma-Aldrich
Potassium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Potassium chloride, AnhydroBeads, −10 mesh, 99.99% trace metals basis
Sigma-Aldrich
Sodium acetate, 99.995% trace metals basis
Sigma-Aldrich
(−)-Epicatechin, ≥98% (HPLC), from green tea
Sigma-Aldrich
Potassium chloride solution, 0.075 M, sterile-filtered, BioXtra, suitable for cell culture
Sigma-Aldrich
Chlorogenic acid, ≥95% (titration)
Sigma-Aldrich
Sodium acetate, anhydrous, Molecular Biology, ≥99%
Sigma-Aldrich
Sodium acetate, powder, BioReagent, suitable for electrophoresis, suitable for cell culture, suitable for insect cell culture, ≥99%
Sigma-Aldrich
Sodium acetate, meets USP testing specifications, anhydrous
Sigma-Aldrich
Sodium acetate, BioXtra, ≥99.0%
Supelco
Potassium chloride solution, ~3 M KCl, saturated with silver chloride
Sigma-Aldrich
Potassium chloride, 99.999% trace metals basis
Sigma-Aldrich
Potassium chloride, BioXtra, ≥99.0%
Sigma-Aldrich
Methanol, Laboratory Reagent, ≥99.6%
Sigma-Aldrich
Sodium acetate, puriss. p.a., ACS reagent, reag. Ph. Eur., anhydrous
Sigma-Aldrich
Sodium acetate, anhydrous, ReagentPlus®, ≥99.0%