Merck
  • Engineered collagen hydrogels for the sustained release of biomolecules and imaging agents: promoting the growth of human gingival cells.
Engineered collagen hydrogels for the sustained release of biomolecules and imaging agents: promoting the growth of human gingival cells.
International journal of nanomedicine (2014-11-28)
Jonghoon Choi, Hoyoung Park, Taeho Kim, Yoon Jeong, Myoung Hwan Oh, Taeghwan Hyeon, Assaf A Gilad, Kwan Hyi Lee
ABSTRACT
We present here the in vitro release profiles of either fluorescently labeled biomolecules or computed tomography contrast nanoagents from engineered collagen hydrogels under physiological conditions. The collagen constructs were designed as potential biocompatible inserts into wounded human gingiva. The collagen hydrogels were fabricated under a variety of conditions in order to optimize the release profile of biomolecules and nanoparticles for the desired duration and amount. The collagen constructs containing biomolecules/nanoconstructs were incubated under physiological conditions (ie, 37°C and 5% CO2) for 24 hours, and the release profile was tuned from 20% to 70% of initially loaded materials by varying the gelation conditions of the collagen constructs. The amounts of released biomolecules and nanoparticles were quantified respectively by measuring the intensity of fluorescence and X-ray scattering. The collagen hydrogel we fabricated may serve as an efficient platform for the controlled release of biomolecules and imaging agents in human gingiva to facilitate the regeneration of oral tissues.
Sigma-Aldrich
Fluorescein isothiocyanate isomer I, suitable for protein labeling, ≥90% (HPLC), powder
Sigma-Aldrich
Fluorescein isothiocyanate isomer I, ≥97.5% (HPLC)
Sigma-Aldrich
Fluorescein 5(6)-isothiocyanate, BioReagent, suitable for fluorescence, mixture of 2 components, ≥90% (HPLC)
Sigma-Aldrich
Fluorescein 5(6)-isothiocyanate, ≥90% (HPLC)
Sigma-Aldrich
(3-Aminopropyl)triethoxysilane, ≥98%
Sigma-Aldrich
(3-Aminopropyl)triethoxysilane, 99%
Sigma-Aldrich
(3-Aminopropyl)triethoxysilane, ≥98.0%
Sigma-Aldrich
(3-Aminopropyl)triethoxysilane, packaged for use in deposition systems, ≥98%
Avanti
18:1 DGS-NTA(Ni), Avanti Research - A Croda Brand
Avanti
14:0 Lyso PC, 1-myristoyl-2-hydroxy-sn-glycero-3-phosphocholine, chloroform
Avanti
14:0 Lyso PC, 1-myristoyl-2-hydroxy-sn-glycero-3-phosphocholine, powder
Avanti
18:0 PEG2000 PE, Avanti Research - A Croda Brand 880120C
Avanti
18:1 DGS-NTA(Ni), Avanti Research - A Croda Brand
Avanti
18:0 PEG2000 PE (Ammonium Salt), Avanti Research - A Croda Brand