Merck
  • In vitro performance of spacers for aerosol delivery during adult mechanical ventilation.
In vitro performance of spacers for aerosol delivery during adult mechanical ventilation.
Journal of aerosol medicine and pulmonary drug delivery (2014-07-23)
Nabile Boukhettala, Thierry Porée, Patrice Diot, Laurent Vecellio
ABSTRACT
During mechanical ventilation, different aerosol generators are employed with various interfaces. The objective of this study was to evaluate the performance of a range of spacers, including a new device called Combihaler® designed for connection with both nebulizers and pressurized Metered-Dose Inhalers (pMDIs). To assess the spacers, we used a ventilator and the Dual Adult Training and Test Lung (model 5600i, Michigan Instruments). Ventilation parameters were measured with and without spacers in volume-controlled and pressure-controlled mode. A filter was placed at the end of the endotracheal tube to measure aerosol delivery. Amikacin (1 g/8 mL) and salbutamol (5 mg/5 mL) were nebulized with an Aeroneb Solo® connected to its T-adapter or the Combihaler® spacer. Salbutamol (100 μg/actuation with 10 actuations) and beclomethasone (250 μg/actuation with 10 actuations) were delivered with a pMDI connected to a Minispacer®, an ACE® spacer, or a Combihaler® spacer. Drug delivery measurements were performed in volume-controlled mode in dry and humidified conditions. Drug deposits on the filter were assayed. The use of spacers and the T-adapter did not change the ventilation parameters (p>0.9). Aerosol delivery of salbutamol and Amikacin by nebulization increased up to three-fold with the Combihaler® compared with the T-adapter in humidified and nonhumidified conditions (p<0.05). Aerosol delivery of salbutamol and beclometasone by pMDI increased up to three-fold with the Combihaler® and the ACE® spacer compared with the Minispacer® in humidified and nonhumidified conditions (p<0.05). Aerosol delivery by pMDIs and vibrating mesh nebulizers using either a T-adapter or spacers was reduced by up to 62.5% in a humidified circuit compared with a nonhumidified circuit. Aerosol delivery via pMDIs and vibrating mesh nebulizers is greater with large spacers (Combihaler® and ACE®) than with smaller spacers (Minispacer®) or a T-adapter, in both humidified and nonhumidified conditions. In humidified conditions, the aerosol delivery decreased with all spacers.
Product Description
Supelco
Water, for TOC analysis
Sigma-Aldrich
Water, BioPerformance Certified
Supelco
Density Standard 998 kg/m3, H&D Fitzgerald Ltd. Quality
Supelco
Water, ACS reagent, for ultratrace analysis
Supelco
Water, suitable for ion chromatography
Sigma-Aldrich
Water, sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Water, for embryo transfer, sterile-filtered, BioXtra, suitable for mouse embryo cell culture
Sigma-Aldrich
Water, Nuclease-Free Water, for Molecular Biology
Sigma-Aldrich
Water, for cell biology, sterile ultrafiltered
Sigma-Aldrich
Water, Molecular Biology, sterile filtered
Sigma-Aldrich
Water, tested according to Ph. Eur.
Supelco
Water, suitable for HPCE, suitable for luminescence, suitable for UV/Vis spectroscopy
Sigma-Aldrich
Sodium Fluoride Solution
Sigma-Aldrich
Water, PCR Reagent
Sigma-Aldrich
E-Toxate Water, endotoxin, free
Pure Water Density Standard, UKAS ISO/IEC17025 and ISO Guide 34 certified, density: 0.9982 g/mL at 20 °C, density: 0.9970 g/mL at 25 °C
Pure Water Density Standard, UKAS ISO/IEC17025 and ISO Guide 34 certified, density: 0.9982 g/mL at 20 °C, density: 0.9970 g/mL at 25 °C
Sigma-Aldrich
Water, ACS reagent
Sigma-Aldrich
Water, Deionized
Sigma-Aldrich
Water, suitable for HPLC
Sigma-Aldrich
Sodium fluoride, 99.99% trace metals basis
Sigma-Aldrich
Water-16O, ≥99.94 atom % 16O
Sigma-Aldrich
Sodium fluoride, anhydrous, powder, 99.99% trace metals basis
Supelco
Salbutamol, VETRANAL®, analytical standard
Supelco
Fluoride ion solution for ISE, 0.1 M F-, for ion-selective electrodes
Sigma-Aldrich
Sodium fluoride, BioXtra, ≥99%
Sigma-Aldrich
Sodium fluoride, BioReagent, suitable for insect cell culture, ≥99%
Sigma-Aldrich
Beclomethasone, ≥99%
Sigma-Aldrich
Salbutamol
Sigma-Aldrich
Sodium fluoride 0.5 M solution