Merck
  • Platinum and palladium nanotubes based on genetically engineered elastin-mimetic fusion protein-fiber templates: synthesis and application in lithium-O₂ batteries.
Platinum and palladium nanotubes based on genetically engineered elastin-mimetic fusion protein-fiber templates: synthesis and application in lithium-O₂ batteries.
Chemistry, an Asian journal (2014-06-26)
Guilue Guo, Thi Hong Anh Truong, Huiteng Tan, Huixiang Ang, Wenyu Zhang, Chen Xu, Xianghong Rui, Zhaolong Hu, Eileen Fong, Qingyu Yan
ABSTRACT
The coupling of proteins with self-assembly properties and proteins that are capable of recognizing and mineralizing specific inorganic species is a promising strategy for the synthesis of nanoscale materials with controllable morphology and functionality. Herein, GPG-AG3 protein fibers with both of these properties were constructed and served as templates for the synthesis of Pt and Pd nanotubes. The protein fibers of assembled GPG-AG3 were more than 10 μm long and had diameters of 20-50 nm. The as-synthesized Pt and Pd nanotubes were composed of dense layers of ~3-5 nm Pt and Pd nanoparticles. When tested as cathodes in lithium-O2 batteries, the porous Pt nanotubes showed low charge potentials of 3.8 V, with round-trip efficiencies of about 65% at a current density of 100 mA g(-1).
Sigma-Aldrich
Palladium(II) chloride solution, 5 wt. % in 10 wt. % HCl
Sigma-Aldrich
Palladium(II) chloride ChemBeads
Sigma-Aldrich
Palladium(II) chloride, anhydrous, 60% Pd basis
Sigma-Aldrich
Palladium(II) chloride, 99.995%
Sigma-Aldrich
Palladium(II) chloride, ≥99.9%
Sigma-Aldrich
Palladium(II) chloride, ReagentPlus®, 99%
Sigma-Aldrich
Palladium(II) chloride, anhydrous, 59-60% palladium (Pd) basis