Merck
  • Quantification of 5,7-dimethoxyflavone in mouse plasma using liquid chromatography-tandem mass spectrometry (LC-MS/MS) and its application to a pharmacokinetic study.
Quantification of 5,7-dimethoxyflavone in mouse plasma using liquid chromatography-tandem mass spectrometry (LC-MS/MS) and its application to a pharmacokinetic study.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences (2014-12-23)
Di Bei, Guohua An
ABSTRACT
5,7-Dimethoxyflavone (5,7-DMF), a natural flavonoid abundant in many plants, has been reported to have many beneficial pharmacological effects, including strong chemopreventive and chemosensitizing properties, anti-oxidant, cardiovascular protectant, and anti-inflammatory activities. However, to date 5,7-DMF was evaluated mainly in vitro and its pharmacokinetics (PK) in vivo remains largely unknown. In addition, current available quantification methods of 5,7-DMF all lack sufficient sensitivity (lower limit of quantification >800 ng/mL). The purposes of our study are to establish a sensitive quantification method of 5,7-DMF using LC-MS/MS and evaluate the PK profile of 5,7-DMF in mouse. Our method was fully validated and all of the fundamental parameters in method validation, including accuracy, precision, sensitivity, selectivity, recovery and stability were evaluated thoroughly in mouse plasma. The calibration curve covered 2-1000 ng/mL with the lower limit of quantification (LLOQ) of 2 ng/mL. The inter-run and intra-run precision and accuracy were less than 15% of nominal concentrations. The matrix effect and recovery yield were within ±15% of nominal concentrations. In the PK study, 5,7-DMF was detectable in mouse plasma up to 21 h, with a terminal half-life of 11.5h. The clearance of 5,7-DMF (CL/F) is 22.3 L/h/kg and area under the curve (AUCinf) is 449 h ng/mL. In conclusion, a fast, accurate, sensitive and selective quantification method of 5,7-DMF was established and the developed method was successfully applied to a PK study of 5,7-DMF following oral administration of 5,7-DMF in mice.
Sigma-Aldrich
Ammonium formate, BioUltra, ≥99.0% (calc. based on dry substance, NT)
Supelco
Ethylene glycol, analytical standard
Sigma-Aldrich
Ethylene glycol, BioUltra, ≥99.5% (GC)
Supelco
Ammonium formate, eluent additive for LC-MS, LiChropur, ≥99.0%
Sigma-Aldrich
Ammonium formate, ≥99.995% trace metals basis
Sigma-Aldrich
Ethylene glycol, anhydrous, 99.8%
Supelco
Ethylene glycol, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Ethylene glycol solution, suitable for NMR (reference standard), 80% in DMSO-d6 (99.9 atom % D), NMR tube size 5 mm × 8 in.
USP
Ethylene glycol, United States Pharmacopeia (USP) Reference Standard
Supelco
Residual Solvent - Acetonitrile(solution in DMSO), Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Ammonium formate, reagent grade, 97%
Sigma-Aldrich
Ethylene glycol, ReagentPlus®, ≥99%
Supelco
Acetonitrile(Neat), Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Ethylene glycol, spectrophotometric grade, ≥99%
Supelco
Ethyl acetate, analytical standard
Supelco
Acetonitrile, analytical standard
Sigma-Aldrich
Formic acid, ≥95%, FCC, FG
Sigma-Aldrich
Ethyl acetate, ≥99%, FCC, FG
Sigma-Aldrich
Ethyl acetate, natural, ≥99%, FCC, FG
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
Sigma-Aldrich
Ethyl acetate, anhydrous, 99.8%
Sigma-Aldrich
Ethyl acetate, ReagentPlus®, ≥99.8%
Sigma-Aldrich
6, 2′, 4′-trimethoxyflavone, ≥98% (HPLC)
Sigma-Aldrich
Acetonitrile, electronic grade, 99.999% trace metals basis
Sigma-Aldrich
Ammonium formate solution, BioUltra, 10 M in H2O
Sigma-Aldrich
Ethylene glycol 5 M solution
USP
Residual Solvent Class 2 - Acetonitrile, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Acetonitrile, biotech. grade, ≥99.93%
Sigma-Aldrich
Acetonitrile, ACS reagent, ≥99.5%
Sigma-Aldrich
Ethyl acetate, biotech. grade, ≥99.8%