Merck
  • Evaluation of two sub-2 μm stationary phases, core-shell and totally porous monodisperse, in the second dimension of on-line comprehensive two dimensional liquid chromatography, a case study: separation of milk peptides after expiration date.
Evaluation of two sub-2 μm stationary phases, core-shell and totally porous monodisperse, in the second dimension of on-line comprehensive two dimensional liquid chromatography, a case study: separation of milk peptides after expiration date.
Journal of chromatography. A (2014-12-17)
Eduardo Sommella, Giacomo Pepe, Giovanni Ventre, Francesco Pagano, Michele Manfra, Giuseppe Pierri, Omar Ismail, Alessia Ciogli, Pietro Campiglia
ABSTRACT
Milk is a rich source of bioactive peptides of great interest for their healthy properties. These peptides are usually encrypted in the sequences of proteins and are released after time dependent proteolysis as very complex hydrolysates. In order to separate and identify the bioactive sequences, we developed an on-line comprehensive two dimensional liquid chromatography approach using the high performance combined with the ultra high performance conditions. A microbore reversed phase (C18 silica, 5 μm) column was employed in first dimension, while, in second dimension, two different UHPLC columns, packed with C18 silica, were tested: a new column based on monodisperse sub-2 μm fully porous particles with high surface area (50 mm × 3.0 mm, 1.9 μm d.p., from Supelco), and a column based on sub-2 μm core-shell particles (50 mm × 3.0 mm, 1.7μm d.p, from Phenomenex(®)). Both set-ups were compared, showing high peak capacity values with respect to a high efficiency monodimensional method, maintaining the same analysis time. Satisfactory selectivity was obtained through the use of different pH between the two dimension, while a very fast continuous shifted gradient in second dimension ensured a good employment of the 2D separation space.
Sigma-Aldrich
Acetonitrile, suitable for DNA synthesis, ≥99.9% (GC)
USP
Residual Solvent Class 2 - Acetonitrile, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Acetonitrile, ACS reagent, ≥99.5%
Sigma-Aldrich
Acetonitrile, ReagentPlus®, 99%
Sigma-Aldrich
Formic acid, ACS reagent, ≥88%
Sigma-Aldrich
Formic acid, ≥95%, FCC, FG
Sigma-Aldrich
Formic acid, ACS reagent, ≥96%
Sigma-Aldrich
Formic acid solution, BioUltra, 1.0 M in H2O
Sigma-Aldrich
Trifluoroacetic acid, ≥99%, for protein sequencing
Supelco
Trifluoroacetic acid, analytical standard
Sigma-Aldrich
Trifluoroacetic acid, ReagentPlus®, 99%
Sigma-Aldrich
Trifluoroacetic acid, suitable for HPLC, ≥99.0%
Supelco
Acetonitrile, analytical standard
Sigma-Aldrich
Acetonitrile, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Acetonitrile, HPLC Plus, ≥99.9%
Sigma-Aldrich
Acetonitrile, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
Sigma-Aldrich
Acetonitrile, biotech. grade, ≥99.93%
Sigma-Aldrich
Acetonitrile, electronic grade, 99.999% trace metals basis
Sigma-Aldrich
Formic acid, reagent grade, ≥95%
Sigma-Aldrich
Trifluoroacetic acid, puriss. p.a., suitable for HPLC, ≥99.0% (GC)
Supelco
Acetonitrile(Neat), Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Residual Solvent - Acetonitrile(solution in DMSO), Pharmaceutical Secondary Standard; Certified Reference Material