Merck
  • Metabolite and peptide levels in plasma and CSF differentiating healthy controls from patients with newly diagnosed Parkinson's disease.
Metabolite and peptide levels in plasma and CSF differentiating healthy controls from patients with newly diagnosed Parkinson's disease.
Journal of Parkinson's disease (2014-06-15)
Miles Trupp, Pär Jonsson, Annika Ohrfelt, Henrik Zetterberg, Ogonna Obudulu, Linus Malm, Anna Wuolikainen, Jan Linder, Thomas Moritz, Kaj Blennow, Henrik Antti, Lars Forsgren
ABSTRACT
Parkinson's disease (PD) is a progressive, multi-focal neurodegenerative disease for which there is no effective disease modifying treatment. A critical requirement for designing successful clinical trials is the development of robust and reproducible biomarkers identifying PD in preclinical stages. To investigate the potential for a cluster of biomarkers visualized with multiple analytical platforms to provide a clinically useful tool. Gas Chromatography-Mass Spectrometry (GC-TOFMS) based metabolomics and immunoassay-based protein/peptide analyses on samples from patients with PD diagnosed in Northern Sweden. Low molecular weight compounds from both plasma and cerebrospinal fluid (CSF) from 20 healthy subjects (controls) and 20 PD patients at the time of diagnosis (baseline) were analyzed. In plasma, we found a significant increase in several amino acids and a decrease in C16-C18 saturated and unsaturated fatty acids in patients as compared to control subjects. We also observed an increase in plasma levels of pyroglutamate and 2-oxoisocaproate (ketoleucine) that may be indicative of increased metabolic stress in patients. In CSF, there was a generally lower level of metabolites in PD as compared to controls, with a specific decrease in 3-hydroxyisovaleric acid, tryptophan and creatinine. Multivariate analysis and modeling of metabolites indicates that while the PD samples can be separated from control samples, the list of detected compounds will need to be expanded in order to define a robust predictive model. CSF biomarker immunoassays of candidate peptide/protein biomarkers revealed a significant decrease in the levels of Aβ-38 and Aβ-42, and an increase in soluble APPα in CSF of patients. Furthermore, these peptides showed significant correlations to each other, and positive correlations to the CSF levels of several 5- and 6-carbon sugars. However, combining these metabolites and proteins/peptides into a single model did not significantly improve the statistical analysis. Together, this metabolomics study has detected significant alterations in plasma and CSF levels of a cluster of amino acids, fatty acids and sugars based on clinical diagnosis and levels of known protein and peptide biomarkers.
Product Description
Supelco
Salicylic acid, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Cholesterol, from sheep wool, ≥92.5% (GC), powder
Sigma-Aldrich
Salicylic acid, ≥99%, FG
Supelco
Cholesterol solution, certified reference material, 10 mg/mL in chloroform
Sigma-Aldrich
SyntheChol® NS0 Supplement, 500 ×, synthetic cholesterol, animal component-free, aqueous solution, sterile-filtered, suitable for cell culture
Sigma-Aldrich
Salicylic acid, ACS reagent, ≥99.0%
Sigma-Aldrich
Myristic acid, natural, ≥98.5%, FG
Supelco
Palmitic acid, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Myristic acid, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Myristic acid, ≥95%, FCC, FG
Sigma-Aldrich
Salicylic acid, meets analytical specification of Ph. Eur., BP, USP, 99.5-100.5% (calc. to the dried substance)
Supelco
Palmitic acid, analytical standard
Sigma-Aldrich
Palmitic acid, BioXtra, ≥99%
Sigma-Aldrich
Cholesterol, from lanolin, ≥99.0% (GC)
Sigma-Aldrich
Chlorotrimethylsilane, puriss., ≥99.0% (GC)
Sigma-Aldrich
Chlorotrimethylsilane, ≥98.0% (GC)
Sigma-Aldrich
Myristic acid, ≥98.0% (GC)
Sigma-Aldrich
Palmitic acid, ≥98% palmitic acid basis (GC)
Supelco
N-Methyl-bis(trifluoroacetamide), derivatization grade (GC derivatization), LiChropur, ≥99.0% (GC)
Sigma-Aldrich
Chlorotrimethylsilane, Wacker Chemie AG, ≥99.0% (GC)
Sigma-Aldrich
Salicylic acid, ReagentPlus®, ≥99%
Sigma-Aldrich
Palmitic acid, ≥98%, FCC, FG
Sigma-Aldrich
Chlorotrimethylsilane, purified by redistillation, ≥99%
Sigma-Aldrich
Cholesterol, tested according to Ph. Eur.
Sigma-Aldrich
Salicylic acid, puriss. p.a., ≥99.0% (T)
Sigma-Aldrich
Salicylic acid, BioXtra, ≥99.0%
Supelco
N-Methyl-bis(trifluoroacetamide), derivatization grade (GC derivatization), LiChropur, ≥97.0% (GC)
Sigma-Aldrich
Cholesterol, Sigma Grade, ≥99%
Sigma-Aldrich
Salicylic acid, suitable for plant cell culture
Sigma-Aldrich
Cholesterol, powder, BioReagent, suitable for cell culture, ≥99%