Merck
  • Polysaccharide nanofibers with variable compliance for directing cell fate.
Polysaccharide nanofibers with variable compliance for directing cell fate.
Journal of biomedical materials research. Part A (2014-05-24)
Xu Jiang, Mui Hoon Nai, Chwee Teck Lim, Catherine Le Visage, Jerry K Y Chan, Sing Yian Chew
ABSTRACT
Cells perceive their microenvironment through physical and mechanical cues, such as extracellular matrix topography or stiffness. In this study, we developed a polysaccharide scaffold that can provide combined substrate topography and matrix compliance signals to direct cell fate. Pullulan/dextran (P/D) nanofibers were fabricated with variable stiffness by in situ crosslinking during electrospinning. By varying the chemical crosslinking content between 10, 12, 14, and 16%, (denoted as STMP10, STMP12, STMP14, and STMP16 respectively), scaffold mechanical stiffness was altered. We characterized substrate stiffness by various methods. Under hydrated conditions, atomic force microscopy and tensile tests of bulk scaffolds were conducted. Under dry conditions, tensile tests of scaffolds and single nanofibers were examined. In addition, we evaluated the efficacy of the scaffolds in directing stem cell differentiation. Using human first trimester mesenchymal stem cells (fMSCs) cultured on STMP14 P/D scaffolds (Young's modulus: 7.84 kPa) in serum-free neuronal differentiation medium exhibited greatest extent of differentiation. Cells showed morphological changes and significantly higher expression of motor neuron markers. Further analyses by western blotting also revealed the enhanced expression of choline acetyltransferase on STMP14 (7.84 kPa) and STMP16 (11.08 kPa) samples as compared to STMP12 (7.19 kPa). Taken together, this study demonstrates that the stiffness of P/D nanofibers can be altered by differential in situ crosslinking during electrospinning and suggests the feasibility of using such polysaccharide nanofibers in supporting fMSC neuronal commitment.
Product Description
USP
Glycine, United States Pharmacopeia (USP) Reference Standard
Glycine, European Pharmacopoeia (EP) Reference Standard
Supelco
Glycine, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Glycine, ReagentPlus®, ≥99% (HPLC)
Sigma-Aldrich
Bicinchoninic acid disodium salt hydrate, ≥98% (HPLC)
Sigma-Aldrich
Sodium dodecyl sulfate, ≥98.0% (GC)
SAFC
Glycine
Sigma-Aldrich
Glycine, ACS reagent, ≥98.5%
Sigma-Aldrich
Sodium hydroxide solution, 1.0 N, BioReagent, suitable for cell culture
Sigma-Aldrich
Sodium hydroxide solution, BioUltra, Molecular Biology, 10 M in H2O
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, Molecular Biology, ≥98.5% (GC), free-flowing, Redi-Dri
Sigma-Aldrich
Sodium dodecyl sulfate, ≥98.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, BioXtra, ≥99.0% (GC)
Sigma-Aldrich
Sodium hydroxide, pellets, semiconductor grade, 99.99% trace metals basis
Sigma-Aldrich
Sodium hydroxide, BioXtra, ≥98% (acidimetric), pellets (anhydrous)
Supelco
Sodium hydroxide solution, 49-51% in water, eluent for IC
Sigma-Aldrich
Sodium hydroxide solution, 50% in H2O
Sigma-Aldrich
Sodium hydroxide, reagent grade, 97%, flakes
Sigma-Aldrich
Sodium hydroxide, BioUltra, suitable for luminescence, ≥98.0% (T), pellets
Sigma-Aldrich
Sodium dodecyl sulfate, 92.5-100.5% based on total alkyl sulfate content basis
Supelco
Sodium dodecyl sulfate, suitable for ion pair chromatography, LiChropur, ≥99.0%
Supelco
Sodium dodecyl sulfate, dust-free pellets, suitable for electrophoresis, Molecular Biology, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, Molecular Biology, ≥98.5% (GC)
Sigma-Aldrich
Sodium hydroxide, ACS reagent, ≥97.0%, pellets
Sigma-Aldrich
Sodium hydroxide, beads, 16-60 mesh, reagent grade, 97%
Sigma-Aldrich
Sodium dodecyl sulfate, ACS reagent, ≥99.0%
Sigma-Aldrich
Sodium hydroxide, reagent grade, ≥98%, pellets (anhydrous)
Sigma-Aldrich
Sodium hydroxide solution, 5.0 M
Sigma-Aldrich
Sodium hydroxide, reagent grade, 97%, powder
Sigma-Aldrich
Sodium hydroxide, puriss., meets analytical specification of Ph. Eur., BP, NF, E524, 98-100.5%, pellets