Merck
  • Fungal metabolite nigerloxin ameliorates diabetic nephropathy and gentamicin-induced renal oxidative stress in experimental rats.
Fungal metabolite nigerloxin ameliorates diabetic nephropathy and gentamicin-induced renal oxidative stress in experimental rats.
Naunyn-Schmiedeberg's archives of pharmacology (2014-06-12)
Bharathinagar S Suresha, Krishnapura Srinivasan
ABSTRACT
Elevated polyol pathway enzyme activities and oxidative stress play an important role in the development and progression of diabetic nephropathy. Here, we investigated the beneficial influence of nigerloxin, a fungal metabolite and a potent aldose reductase inhibitor and free radical scavenger in the kidney of streptozotocin-induced diabetic rats. A group of diabetic rats was orally administered with nigerloxin for 30 days (100 mg/kg). Diabetic rats showed increased lipid peroxides, advanced glycation end products (AGEs), elevated activities of polyol pathway enzymes, and lowered antioxidant defense system in kidney. Administration of nigerloxin decreased kidney lipid peroxides and AGEs. Activities of polyol pathway enzymes were reduced while activities of all antioxidant enzymes, glutathione, and ascorbic acid were elevated in the kidney of nigerloxin-treated diabetic rats. We also investigated antioxidant potential of nigerloxin in gentamicin-induced nephrotoxicity in Wistar rats. Groups of rats were orally administered with nigerloxin for 8 days (25 mg or 100 mg kg(-1) body weight day(-1)) along with gentamicin (80 mg/kg, i.p., for 8 days). Gentamicin induced increase in lipid peroxides, decrease in glutathione and activities of antioxidant enzymes in the kidney, and increase in blood creatinine, and urea concentrations were significantly countered by nigerloxin treatment. Thus, the results indicated the beneficial influence of nigerloxin on polyol pathway and oxidative stress associated with diabetes, which are implicated in ameliorating the development of diabetic nephropathy. Nigerloxin also ameliorated oxidative stress induced by gentamicin in the renal tissue.
Product Description
Supelco
Fructose, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Guaiacol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
D-(−)-Fructose, ≥99% (HPLC), BioReagent, suitable for cell culture, suitable for insect cell culture
Sigma-Aldrich
D-(−)-Fructose, ≥99% (HPLC), BioXtra
Sigma-Aldrich
L-Ascorbic acid, FCC, FG
Sigma-Aldrich
1-Chloro-2,4-dinitrobenzene, ≥99%
Sigma-Aldrich
Guaiacol, natural, ≥99%, FG
Supelco
L-Ascorbic acid, analytical standard
Millipore
Hydrogen peroxide solution, 3%, suitable for microbiology
Sigma-Aldrich
L-Ascorbic acid, puriss. p.a., ≥99.0% (RT)
Sigma-Aldrich
L-Ascorbic acid, BioUltra, ≥99.5% (RT)
Millipore
D-(−)-Fructose, ≥99.0% (HPLC), suitable for microbiology
Sigma-Aldrich
D-(−)-Fructose, BioUltra, ≥99.0% (HPLC)
Sigma-Aldrich
L-Ascorbic acid, tested according to Ph. Eur.
Sigma-Aldrich
L-Ascorbic acid, puriss. p.a., ACS reagent, reag. ISO, Ph. Eur., 99.7-100.5% (oxidimetric)
Supelco
Hydrogen peroxide solution, ≥30%, for trace analysis
Sigma-Aldrich
D-(−)-Fructose, tested according to Ph. Eur.
Sigma-Aldrich
L-Ascorbic acid, 99%
Sigma-Aldrich
Hydrogen peroxide solution, tested according to Ph. Eur.
Sigma-Aldrich
Urea-12C, 99.9 atom % 12C
Sigma-Aldrich
1-Chloro-2,4-dinitrobenzene, 97%
Sigma-Aldrich
tert-Butyl hydroperoxide solution, 5.0-6.0 M in decane
Sigma-Aldrich
5,5′-Dithiobis(2-nitrobenzoic acid), ReagentPlus®, 99%
Sigma-Aldrich
tert-Butyl hydroperoxide solution, 70 wt. % in H2O
Sigma-Aldrich
tert-Butyl hydroperoxide solution, 5.0-6.0 M in nonane
Sigma-Aldrich
DL-Glyceraldehyde, ≥90% (GC)
Supelco
D-(−)-Fructose, analytical standard, analytical standard for fructose assay kit, for use with enzymatic assay kit FA20
Sigma-Aldrich
D-(−)-Fructose, ≥99% (HPLC)
Sigma-Aldrich
L-Glutathione oxidized disodium salt, suitable for cell culture, BioReagent
Sigma-Aldrich
L-Ascorbic acid, suitable for cell culture, suitable for plant cell culture, ≥98%