Merck
  • Polymeric micelles with citraconic amide as pH-sensitive bond in backbone for anticancer drug delivery.
Polymeric micelles with citraconic amide as pH-sensitive bond in backbone for anticancer drug delivery.
International journal of pharmaceutics (2014-05-20)
Jun Cao, Ting Su, Longgui Zhang, Rong Liu, Gang Wang, Bin He, Zhongwei Gu
ABSTRACT
A novel pH-sensitive polymeric micelle was reported. Methoxy poly(ethylene glycol)-b-poly(ϵ-caprolactone) copolymer with citraconic amide as pH-sensitive bond was synthesized (mPEG-pH-PCL). The copolymers self-assembled into micelles to encapsulate anticancer drug doxorubicin (DOX). The morphology, size and size distribution, drug release profile and in vitro anticancer activity of the DOX loaded mPEG-pH-PCL micelles were studied. The results showed that the mean size of the micelles was around 120 nm, the drug loading content and encapsulation efficiency of the mPEG-pH-PCL micelles were 6.8% and 54.3%, respectively. The mean diameter and size distribution of the mPEG-pH-PCL micelles increased significantly when soaking in medium with pH 5.5. The drug release of micelles in pH 5.5 was much faster than that in pH 7.4. The confocal laser microscopy and flow cytometry measurements indicated that the weak acidity of endosomes broke the citraconic amide bonds in the copolymer backbones and triggered the fast release of DOX. The in vitro IC50 of the drug loaded mPEG-pH-PCL micelles was lower than that of drug loaded polymeric micelles without pH-sensitivity to both HepG2 and 4T1 cancer cells.
Product Description
USP
Ethylene glycol, United States Pharmacopeia (USP) Reference Standard
Supelco
Ethylene glycol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Potassium chloride, tested according to Ph. Eur.
Supelco
Potassium chloride solution, conductance standard A acc. to ISO 7888, 0.1 M KCl
Supelco
Potassium chloride solution, conductance standard C acc. to ISO 7888, 0.001 M KCl
Supelco
ISA (ionic strength adjustment solution: 1 M KCl), 1 M KCl
Supelco
Potassium chloride solution, conductance standard B acc. to ISO 7888, 0.01 M KCl
Sigma-Aldrich
Chloroform-d, "100%", 99.96 atom % D, contains 0.03 % (v/v) TMS
Sigma-Aldrich
Chloroform-d, 99.8 atom % D
Sigma-Aldrich
Chloroform-d, "100%", 99.95 atom % D
Sigma-Aldrich
Chloroform-d, 99.8 atom % D
Sigma-Aldrich
Chloroform-d, ≥99.8 atom % D, anhydrous
Sigma-Aldrich
Chloroform-d, 99.8 atom % D, contains 0.05 % (v/v) TMS
Sigma-Aldrich
Chloroform-d, 99.8 atom % D, contains 0.03 % (v/v) TMS
Sigma-Aldrich
Chloroform-d, ≥99.8 atom % D, contains 0.5 wt. % silver foil as stabilizer
Sigma-Aldrich
Potassium chloride, ≥99.99% trace metals basis
Sigma-Aldrich
Chloroform-d, "100%", 99.96 atom % D, contains 0.5 wt. % silver wire as stabilizer
Sigma-Aldrich
Ethylene glycol, BioUltra, ≥99.5% (GC)
Supelco
Ethylene glycol, analytical standard
Sigma-Aldrich
Potassium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Sigma-Aldrich
Chloroform-d, ≥99.8 atom % D, contains 0.5 wt. % silver foil as stabilizer, 0.03 % (v/v) TMS
Sigma-Aldrich
Potassium chloride, AnhydroBeads, −10 mesh, 99.99% trace metals basis
Sigma-Aldrich
Potassium chloride solution, 0.075 M, sterile-filtered, BioXtra, suitable for cell culture
Supelco
Potassium chloride solution, ~3 M KCl, saturated with silver chloride
Sigma-Aldrich
Potassium chloride, 99.999% trace metals basis
Sigma-Aldrich
Potassium chloride solution, BioUltra, Molecular Biology, ~1 M in H2O
Sigma-Aldrich
Potassium chloride, BioXtra, ≥99.0%
Sigma-Aldrich
Potassium chloride, BioUltra, Molecular Biology, ≥99.5% (AT)
Sigma-Aldrich
Potassium chloride, ReagentPlus®, ≥99.0%
Sigma-Aldrich
Potassium chloride, Molecular Biology, ≥99.0%