Merck
  • Neuroprotection against degeneration of sk-N-mc cells using neuron growth factor-encapsulated liposomes with surface cereport and transferrin.
Neuroprotection against degeneration of sk-N-mc cells using neuron growth factor-encapsulated liposomes with surface cereport and transferrin.
Journal of pharmaceutical sciences (2014-07-22)
Yung-Chih Kuo, Pei-Ru Chou
ABSTRACT
Liposomes with Cereport (RMP-7) and transferrin (Tf) (RMP-7/Tf/liposomes) were employed to target the blood-brain barrier (BBB) and to inhibit the degeneration of neurons insulted with fibrillar β-amyloid peptide 1-42 (Aβ1-42). Neuron growth factor (NGF)-encapsulated RMP-7/Tf/liposomes (RMP-7/Tf/NGF-liposomes) were used to permeate a monolayer of human brain-microvascular endothelial cells (HBMECs) regulated by human astrocytes (HAs) and to treat Aβ1-42 -attacked SK-N-MC cells. An increase in RMT-7 concentration increased the particle size, zeta potential, propidium iodide (PI) permeability, and NGF permeability, but decreased the cross-linking efficiency of RMT-7, viability of HBMECs and HAs, and transendothelial electrical resistance (TEER). In addition, an increase in Tf concentration enhanced the particle size, viability of HBMECs, HAs, and SK-N-MC cells, PI permeability, and NGF permeability, but reduced the zeta potential, cross-linking efficiency of RMT-7 and Tf, and TEER. RMP-7/Tf/NGF-liposomes can transport NGF across the BBB and improve the neuroprotection for Alzheimer's disease therapy in preclinical trials.
Sigma-Aldrich
SyntheChol® NS0 Supplement, 500 ×, synthetic cholesterol, animal component-free, sterile-filtered, aqueous solution, suitable for cell culture
Supelco
Chloroform, Pharmaceutical Secondary Standard; Certified Reference Material
SAFC
Cholesterol, from sheep wool, Controlled origin, meets USP/NF testing specifications
Sigma-Aldrich
Cholesterol, from lanolin, ≥99.0% (GC)
Sigma-Aldrich
Propidium iodide, ≥94% (HPLC)
Sigma-Aldrich
Chloroform, anhydrous, contains amylenes as stabilizer, ≥99%
Sigma-Aldrich
Chloroform, anhydrous, ≥99%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
2-Phenylindole, technical grade, 95%
Sigma-Aldrich
Fura 2-AM, BioReagent, suitable for fluorescence, ≥95.0% (HPLC)
Sigma-Aldrich
HEPES, BioUltra, Molecular Biology, ≥99.5% (T)
Sigma-Aldrich
Cholesterol, tested according to Ph. Eur.
Sigma-Aldrich
Sodium pyruvate, BioXtra, ≥99%
Sigma-Aldrich
Cholesterol, Sigma Grade, ≥99%
Sigma-Aldrich
Sodium pyruvate, ReagentPlus®, ≥99%
Sigma-Aldrich
Cholesterol, powder, BioReagent, suitable for cell culture, ≥99%
Sigma-Aldrich
Sodium pyruvate, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥99%
Sigma-Aldrich
HEPES, BioXtra, suitable for mouse embryo cell culture, ≥99.5% (titration)
Sigma-Aldrich
HEPES, ≥99.5% (titration)
Sigma-Aldrich
HEPES, BioXtra, pH 5.0-6.5 (1 M in H2O), ≥99.5% (titration)
Sigma-Aldrich
HEPES, BioPerformance Certified, ≥99.5% (titration), suitable for cell culture
Sigma-Aldrich
Fura 2-AM, ≥95% (HPLC)
Sigma-Aldrich
Propidium iodide, ≥94.0% (HPLC)
Sigma-Aldrich
Sodium pyruvate, powder, BioXtra, suitable for mouse embryo cell culture
Sigma-Aldrich
Sodium pyruvate, Hybri-Max, powder, suitable for hybridoma
Sigma-Aldrich
Cholesterol, from sheep wool, ≥92.5% (GC), powder
Sigma-Aldrich
Chloroform, ≥99%, PCR Reagent, contains amylenes as stabilizer
Sigma-Aldrich
Chloroform, biotech. grade, ≥99.8%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Chloroform, contains amylenes as stabilizer, ACS reagent, ≥99.8%
Sigma-Aldrich
Chloroform, contains ethanol as stabilizer, meets analytical specification of BP, 99-99.4% (GC)
Supelco
Chloroform, analytical standard