Merck
  • MiR-126-5p regulates osteolysis formation and stromal cell proliferation in giant cell tumor through inhibition of PTHrP.
MiR-126-5p regulates osteolysis formation and stromal cell proliferation in giant cell tumor through inhibition of PTHrP.
Bone (2014-06-29)
Wang Zhou, Huabin Yin, Ting Wang, Tielong Liu, Zhenxi Li, Wangjun Yan, Dianwen Song, Haiyan Chen, Jia Chen, Wei Xu, Xinghai Yang, Zhipeng Wu, Jianru Xiao
ABSTRACT
Parathyroid hormone-related protein (PTHrP) has been identified to play a crucial role in osteolysis formation and stromal cell (GCTSC) proliferation in giant cell tumor (GCT). MiR-126-5p is an intronic miRNA identified as tumor suppressor in many tumors, but its role in GCT is poorly understood. We found that miR-126-5p was decreased in GCT and could directly regulate PTHrP expression. Furthermore, miR-126-5p could control osteoclast (OC) differentiation, GCTSC proliferation and osteolysis formation in GCT through negative regulation of PTHrP. Thus, these results suggest that miR-126-5p could directly target PTHrP and have a tumor suppressor function in GCT.
Sigma-Aldrich
Ser-Phe-Leu-Leu-Arg-Asn-Pro-Asn-Asp-Lys-Tyr-Glu-Pro-Phe, ≥97% (HPLC)
Digoxigenin, European Pharmacopoeia (EP) Reference Standard
Supelco
Digoxigenin, analytical standard