Merck
  • Development of biodegradable nanoparticles for liver-specific ribavirin delivery.
Development of biodegradable nanoparticles for liver-specific ribavirin delivery.
Journal of pharmaceutical sciences (2014-10-23)
Tsutomu Ishihara, Kohei Kaneko, Tomoaki Ishihara, Tohru Mizushima
ABSTRACT
Ribavirin is an antiviral drug used for the treatment of chronic hepatitis C. However, ribavirin induces severe side effects such as hemolytic anemia. In this study, we prepared biodegradable nanoparticles as ribavirin carriers to modulate the pharmacokinetics of the drug. The nanoparticles encapsulating ribavirin monophosphate (RMP) were prepared from the blend of poly(d,l-lactic acid) homopolymer and arabinogalactan (AG)-poly(l-lysine) conjugate by using the solvent diffusion method in the presence of iron (III). RMP was efficiently and stably embedded in the nanoparticles and gradually released for 37 days in phosphate-buffered saline at 37°C. The coating of AG on the nanoparticles surfaces was verified by measuring the zeta potentials and performing an aggregation test of the nanoparticles using galactose-binding lectin. Moreover, the nanoparticles were efficiently internalized in cultured HepG2 cells. Ribavirin was drastically accumulated to the liver of mice after intravenous administration of the RMP-loaded nanoparticles, after which the ribavirin content gradually decreased for at least 7 days. Our results indicated successful development of nanoparticles with dual functions, targeting to the liver and sustained release of ribavirin, and suggested that the present strategy could help to advance the clinical application of ribavirin as a therapeutic agent for chronic hepatitis C.
Product Description
Supelco
Benzocaine, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Sodium sulfate, ≥99.99% trace metals basis
Sigma-Aldrich
Sodium sulfate, tested according to Ph. Eur., anhydrous
Sigma-Aldrich
Sodium sulfate, BioUltra, anhydrous, ≥99.0% (T)
Sigma-Aldrich
Benzocaine, ≥99% (HPLC)
Sigma-Aldrich
Sodium sulfate, BioXtra, ≥99.0%
Sigma-Aldrich
Sodium sulfate, ≥99.0%, suitable for plant cell culture
Sigma-Aldrich
Bicinchoninic acid disodium salt hydrate, ≥98% (HPLC)
Sigma-Aldrich
1,2-Dilinoleoyl-3-palmitoyl-rac-glycerol, ≥95% (TLC), liquid
Sigma-Aldrich
Sodium sulfate, anhydrous, free-flowing, Redi-Dri, ACS reagent, ≥99%
Supelco
Acetonitrile(Neat), Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Residual Solvent - Acetonitrile(solution in DMSO), Pharmaceutical Secondary Standard; Certified Reference Material
USP
Benzocaine, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Sodium sulfate, ReagentPlus®, ≥99.0%
Sigma-Aldrich
Sodium sulfate, anhydrous, free-flowing, Redi-Dri, ReagentPlus®, ≥99%
Benzocaine, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Sodium sulfate, ACS reagent, ≥99.0%, anhydrous, granular
Sigma-Aldrich
Sodium sulfate, ACS reagent, ≥99.0%, anhydrous, powder
Sigma-Aldrich
Trifluoroacetic acid, ReagentPlus®, 99%
Sigma-Aldrich
Acetonitrile, ReagentPlus®, 99%
Sigma-Aldrich
Potassium phosphate monobasic, ACS reagent, ≥99.0%
Sigma-Aldrich
Ultrapure Acetonitrile
Ribavirin, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Acetonitrile, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Trifluoroacetic acid, suitable for HPLC, ≥99.0%
Sigma-Aldrich
Acetonitrile, HPLC Plus, ≥99.9%
Sigma-Aldrich
Acetonitrile, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Hydrogen bromide solution, 33 wt. % in acetic acid
Sigma-Aldrich
Hydrobromic acid, reagent grade, 48%
Sigma-Aldrich
Trifluoroacetic acid, puriss. p.a., suitable for HPLC, ≥99.0% (GC)