Merck
  • Merging bioactivity with liquid chromatography-mass spectrometry-based chemometrics to identify minor immunomodulatory compounds from a Micronesian adaptogen, Phaleria nisidai.
Merging bioactivity with liquid chromatography-mass spectrometry-based chemometrics to identify minor immunomodulatory compounds from a Micronesian adaptogen, Phaleria nisidai.
Journal of chromatography. A (2014-09-15)
Daniel M Kulakowski, Shi-Biao Wu, Michael J Balick, Edward J Kennelly
ABSTRACT
This study presents a strategy based on repeatable reversed-phase LC-TOF-MS methods and chemometric statistical tools, including untargeted PCA and supervised OPLS-DA models, to identify low-yielding compounds with potent immunostimulant activity in Phaleria nisidai (Thymelaeaceae), a plant with a history of use as an adaptogen on the islands of Palau in Micronesia. IFNγ ELISA assays were used to classify chromatographic fractions according to imunomodulatory activity prior to LC-TOF-MS chemometric analysis to target and identify compounds likely to contribute to observed activity. Simplexin, a daphnane diterpene ester, was identified for the first time from this genus and caused an increase in the production of cytokines (IFNγ, IL1β, IL6, and IL13) by peripheral blood mononuclear cells. Five other daphnane diterpene esters were tentatively identified for the first time from this plant based on mass spectral data and are marker metabolites distinguishing active from inactive fractions. This analytical approach increased the efficiency of bioactivity-guided fractionation and has the potential to minimize redundant isolation and identify minor constituents with potent activity from a complex matrix.
Sigma-Aldrich
Isopropyl alcohol, ≥99.7%, FCC, FG
Leucine, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
2-Propanol, anhydrous, 99.5%
Supelco
2-Propanol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
2-Propanol, BioReagent, ≥99.5%, Molecular Biology
Sigma-Aldrich
DL-Leucine, ≥99% (HPLC)
Sigma-Aldrich
2-Propanol, BioUltra, Molecular Biology, ≥99.5% (GC)
Supelco
2-Propanol, analytical standard
Sigma-Aldrich
2-Propanol, HPLC Plus, for HPLC, GC, and residue analysis, 99.9%, poly coated bottles
Supelco
Hexane, analytical standard
USP
2-Propanol, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Isopropyl alcohol, meets USP testing specifications
Sigma-Aldrich
2-Propanol, ACS reagent, ≥99.5%
Sigma-Aldrich
Hexane, Laboratory Reagent, ≥95%
Sigma-Aldrich
Hexane, ReagentPlus®, ≥99%
Sigma-Aldrich
2-Propanol, Laboratory Reagent, ≥99.5%
Sigma-Aldrich
2-Propanol, puriss. p.a., ACS reagent, ≥99.8% (GC)
Sigma-Aldrich
2-Propanol, electronic grade, 99.999% trace metals basis
Sigma-Aldrich
2-Propanol, suitable for HPLC, 99.9%
Sigma-Aldrich
Hexane, suitable for HPLC, ≥95%
Sigma-Aldrich
2-Propanol, HPLC Plus, for HPLC, GC, and residue analysis, 99.9%
Sigma-Aldrich
Hexane, HPLC Plus, for HPLC, GC, and residue analysis, ≥95%
Sigma-Aldrich
2-Propanol, suitable for HPLC, 99.5%