Merck
  • Erbin interacts with c-Cbl and promotes tumourigenesis and tumour growth in colorectal cancer by preventing c-Cbl-mediated ubiquitination and down-regulation of EGFR.
Erbin interacts with c-Cbl and promotes tumourigenesis and tumour growth in colorectal cancer by preventing c-Cbl-mediated ubiquitination and down-regulation of EGFR.
The Journal of pathology (2014-12-19)
Su Yao, Ping Zheng, Hua Wu, Li-Ming Song, Xiao-Fang Ying, Cheng Xing, Ying Li, Zheng-Quan Xiao, Xing-Ni Zhou, Tong Shen, Lin Chen, Yu-Hong Liu, Mao-De Lai, Lin Mei, Tian-Ming Gao, Jian-Ming Li
ABSTRACT
The epidermal growth factor receptor (EGFR) is implicated in many types of cancer, including colorectal cancer (CRC), and has become one of the most common candidates for targeted therapy. Here, we found that Erbin, a member of the leucine-rich repeat and PDZ domain (LAP) family, plays a key role in EGFR signalling. Erbin inhibited EGFR ubiquitination and stabilized the EGFR protein by interacting with c-Cbl. Moreover, the PDZ domain of Erbin was critical for the interaction between Erbin and c-Cbl and EGFR ubiquitination. Interestingly, Erbin expression was elevated in tumour samples from CRC patients, increased in advanced clinical stage disease and correlated with EGFR expression. In vivo studies using mouse xenograft models of CRC showed that Erbin promotes tumour growth, and that the effects of Erbin on tumour growth are mainly related to the regulatory effects of Erbin on EGFR. The azoxymethane (AOM)-induced colon carcinogenesis model in Erbin(ΔC) (/) (ΔC) mice, with the PDZ domain of Erbin deleted, demonstrated that the PDZ domain of Erbin and its regulation of EGFR signalling are necessary for the tumourigenesis and tumour growth of CRC. We found that Erbin promotes tumourigenesis and tumour growth in CRC by stabilizing EGFR. Our study sheds light on developing Erbin, especially its PDZ domain, as a potential target for CRC treatment.
Product Description
Sodium laurilsulfate, European Pharmacopoeia (EP) Reference Standard
SAFC
Sodium deoxycholate
Sigma-Aldrich
Sodium deoxycholate, ≥97% (titration)
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, 20% in H2O
Sigma-Aldrich
Sodium deoxycholate, BioXtra, ≥98.0% (dry matter, NT)
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, Molecular Biology, 10% in H2O
Sigma-Aldrich
Sodium chloride solution, 0.85%
Sigma-Aldrich
Sodium dodecyl sulfate, ≥99.0% (GC), dust-free pellets
Sigma-Aldrich
Sodium chloride, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride, random crystals, 99.9% trace metals basis
Sigma-Aldrich
Sodium chloride-35Cl, 99 atom % 35Cl
Sigma-Aldrich
Sodium dodecyl sulfate, ACS reagent, ≥99.0%
Supelco
Sodium dodecyl sulfate, suitable for ion pair chromatography, LiChropur, ≥99.0%
Sigma-Aldrich
Sodium chloride, BioUltra, Molecular Biology, ≥99.5% (AT)
Sigma-Aldrich
Sodium dodecyl sulfate, ≥98.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, BioUltra, Molecular Biology, ≥99.0% (GC)
Sigma-Aldrich
Sodium chloride, tested according to Ph. Eur.
Sigma-Aldrich
Sodium chloride solution, BioUltra, Molecular Biology, ~5 M in H2O
Supelco
Sodium chloride, reference material for titrimetry, certified by BAM, >99.5%
Sigma-Aldrich
Sodium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Sigma-Aldrich
Sodium dodecyl sulfate, tested according to NF, mixture of sodium alkyl sulfates consisting mainly of sodium dodecyl sulfate
Sigma-Aldrich
Sodium dodecyl sulfate, ≥98.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, Molecular Biology, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ReagentPlus®, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, BioXtra, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, 92.5-100.5% based on total alkyl sulfate content basis
Sigma-Aldrich
Sodium chloride, Molecular Biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Sodium chloride, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Sigma-Aldrich
Sodium chloride, tablet
Sigma-Aldrich
Sodium chloride, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%