Merck
  • A green one-pot synthesis of Pt/TiO2/Graphene composites and its electro-photo-synergistic catalytic properties for methanol oxidation.
A green one-pot synthesis of Pt/TiO2/Graphene composites and its electro-photo-synergistic catalytic properties for methanol oxidation.
Journal of colloid and interface science (2014-08-16)
Lingting Ye, Zhongshui Li, Lian Zhang, Fengling Lei, Shen Lin
ABSTRACT
A facile and green one-pot method was used to synthesize Pt/TiO2/Graphene composites with ethanol as a reducing agent under microwave irradiation. The as-prepared composites were characterized by SEM, TEM, EDX, XPS, XRD and Raman. Electrocatalytic performance of the Pt/TiO2/GNs composites was investigated by cyclic voltammetry (CV), chronoamperometric (CA), COad stripping voltammetry and electrochemical impedance spectrum (EIS). All experimental data have revealed that TiO2 (P25) not only enhanced the reduction ability of ethanol under microwave irradiation but also promoted Pt heterogeneous nucleation to form Pt nanoclusters which are around P25 and loaded on graphene nanosheets (GNs) surface. Electrochemical experiments showed that Pt/TiO2/GNs had much higher catalytic activity and stability toward methanol oxidation reaction (MOR) and better resistance to CO poisoning compared with Pt/GNs and the commercially available Johnson Matthey 20% Pt/C catalyst (Pt/C-JM). Especially under UV irradiation with 20min, Pt/TiO2/GNs composites showed an ultrahigh forward peak current density of 1354mAmg(-1), nearly 2.5 times higher than that of Pt/C-JM, which indicated that the electrocatalytic and photocatalytic properties of Pt/TiO2/GNs had been integrated to boost the catalytic performance for MOR.
Product Description
Supelco
Dehydrated Alcohol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Methanol, suitable for NMR (reference standard)
Supelco
Ethanol solution, certified reference material, 2000 μg/mL in methanol
Supelco
Sulfuric acid, for the determination of nitrogen, ≥97.0%
Supelco
Methanol, analytical standard
Sigma-Aldrich
Phosphorus pentoxide, puriss. p.a., ACS reagent, ≥98.0% (T)
Sigma-Aldrich
Phosphorus pentoxide, desiccant, with moisture indicator
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Phosphorus pentoxide, ReagentPlus®, 99%
Supelco
Phosphorus pentoxide, ≥99.99% trace metals basis
Sigma-Aldrich
Sulfuric acid, 99.999%
Sigma-Aldrich
Phosphorus pentoxide, powder, ACS reagent, ≥98.0%
USP
Dehydrated Alcohol, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Sulfuric acid, puriss., meets analytical specification of Ph. Eur., BP, 95-97%
Sigma-Aldrich
Phosphorus pentoxide, anhydrous, free-flowing, Redi-Dri, ReagentPlus®, 99%
Sigma-Aldrich
Methanol, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, suitable as ACS-grade LC reagent, ≥99.9%
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%
Sigma-Aldrich
Methanol, Laboratory Reagent, ≥99.6%
Supelco
Methanol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Methanol, ACS spectrophotometric grade, ≥99.9%
Sigma-Aldrich
Methanol, Absolute - Acetone free
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Sulfuric acid, ACS reagent, 95.0-98.0%
Sigma-Aldrich
Ethyl alcohol, Pure 190 proof, for molecular biology
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, BioReagent, ≥99.93%
Supelco
Sulfuric acid concentrate, 0.1 M H2SO4 in water (0.2N), eluent concentrate for IC
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, anhydrous, ≥99.5%