Merck
  • A two-enzyme immobilization approach using carbon nanotubes/silica as support.
A two-enzyme immobilization approach using carbon nanotubes/silica as support.
Biotechnology progress (2014-11-08)
Kun Du, Jian Sun, Xiaoyu Zhou, Wei Feng, Xia Jiang, Peijun Ji
ABSTRACT
Multiple enzyme mixtures are attractive for the production of many compounds at an industrial level. We report a practical and novel approach for coimmobilization of two enzymes. The system consists of a silica microsphere core coated with two layers of individually immobilized enzymes. The model enzymes α-amylase (AA) and glucoamylase (GluA) were individually immobilized on carbon nanotubes (CNTs). A CNT-GluA layer was formed by adsorbing CNT-GluA onto silica microsphere. A sol-gel layer with entrapped CNT-AA was then formed outside the CNT-GluA/silica microsphere conjugate. The coimmobilized α-amylase and glucoamylase exhibited 95.1% of the activity of the mixture of free α-amylase and glucoamylase. The consecutive use exhibited a good stability of the coimmobilized enzymes. The developed approach demonstrates advantages, including controlling the ratio of coimmobilized enzymes in an easy way, facilitating diffusion of small molecules in and out of the matrix, and preventing the leaching of enzymes.
Product Description
USP
Sorbitol, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Sulfuric acid, ACS reagent, 95.0-98.0%
Sigma-Aldrich
D-Sorbitol, FCC, FG
Sigma-Aldrich
D-Sorbitol, liquid, tested according to Ph. Eur.
Sigma-Aldrich
D-Sorbitol, 99% (GC)
Supelco
Sorbitol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Bicinchoninic acid disodium salt hydrate, ≥98% (HPLC)
Sigma-Aldrich
D-Sorbitol, ≥98% (GC)
Sigma-Aldrich
D-Sorbitol, ≥98% (GC), Molecular Biology
Sigma-Aldrich
D-Sorbitol, ≥98% (GC), BioReagent, suitable for cell culture, suitable for plant cell culture
Sigma-Aldrich
Tetraethyl orthosilicate, ≥99.0% (GC)
Supelco
Sulfuric acid, for the determination of nitrogen, ≥97.0%
Sigma-Aldrich
D-Sorbitol, BioUltra, ≥99.0% (HPLC)
Sigma-Aldrich
Sulfuric acid, 99.999%
Sigma-Aldrich
Sorbitol F solution, 70 wt. % in H2O, Contains mainly D-sorbitol with lesser amounts of other hydrogenated oligosaccharides
Sigma-Aldrich
Tetraethyl orthosilicate, 99.999% trace metals basis
Sigma-Aldrich
Tetraethyl orthosilicate, reagent grade, 98%
Sigma-Aldrich
D-Sorbitol, ≥98% (GC), BioXtra
Sorbitol, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Nitric-14N acid solution, ~10 N in H2O, 99.99 atom % 14N
Sigma-Aldrich
Tetraethyl orthosilicate, packaged for use in deposition systems
Sigma-Aldrich
Sulfuric acid, puriss., meets analytical specification of Ph. Eur., BP, 95-97%
Supelco
Nitric acid concentrate, 0.1 M HNO3 in water (0.1N), eluent concentrate for IC
Sigma-Aldrich
1,2-Dichloroethane, anhydrous, 99.8%
Sigma-Aldrich
Nitric acid, 70%, purified by redistillation, ≥99.999% trace metals basis
Sigma-Aldrich
Nitric acid, ACS reagent, ≥90.0%
Sigma-Aldrich
MES solution, BioUltra, Molecular Biology, 0.5 M in H2O
Sigma-Aldrich
N-Hydroxysuccinimide, purum, ≥97.0% (T)
Sigma-Aldrich
N-Hydroxysuccinimide, 98%
Supelco
Sulfuric acid concentrate, 0.1 M H2SO4 in water (0.2N), eluent concentrate for IC