Merck
Allelochemicals in the rhizosphere soil of Euphorbia himalayensis.
Journal of agricultural and food chemistry (2014-08-05)
Quan Liu, Dengxue Lu, Hui Jin, Zhiqiang Yan, Xiuzhuang Li, Xiaoyan Yang, Hongru Guo, Bo Qin
ABSTRACT
Weed infestation has been known to cause considerable reductions in crop yields, thereby hindering sustainable agriculture. Many plants in genus Euphorbia affect neighboring plants and other organisms by releasing chemicals into the environment. In view of the serious threat of weeds to agriculture, the allelochemicals of Euphorbia himalayensis and their allelopathic effects were investigated. The extract of root exudates from rhizosphere soil exhibited allelopathic activities against crops (wheat, rape, and lettuce) and grasses (Poa annua, Festuca rubra, and red clover). Bioassay-guided fractionation and isolation from the root extract of E. himalayensis led to the characterization of two ellagic acid derivatives and a jatrophane diterpene, which observably showed phytotoxic activities against lettuce, Festuca arundinacea, and F. rubra. They were further confirmed by ultra-performance liquid chromatography-tandem mass spectrometry to have concentrations of 3.6, 3.8, and 8.99 nmol/g in the rhizospere soil, respectively. Bioassay indicated that the combination of the allelochemicals could be selective plant growth regulator in agriculture.
Sigma-Aldrich
Acetonitrile, electronic grade, 99.999% trace metals basis
Sigma-Aldrich
Methanol solution, suitable for NMR (reference standard), 4% in methanol-d4 (99.8 atom % D), NMR tube size 3 mm × 8 in.
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
Supelco
Acetonitrile, analytical standard
Sigma-Aldrich
Acetonitrile, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Acetonitrile, suitable for DNA synthesis, ≥99.9% (GC)
Sigma-Aldrich
Acetonitrile, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Acetonitrile, HPLC Plus, ≥99.9%
Sigma-Aldrich
Acetonitrile, ReagentPlus®, 99%
Sigma-Aldrich
Acetonitrile, ACS reagent, ≥99.5%
Sigma-Aldrich
Acetonitrile, biotech. grade, ≥99.93%
USP
Residual Solvent Class 2 - Acetonitrile, United States Pharmacopeia (USP) Reference Standard
USP
Methyl alcohol, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Methanol, suitable for NMR (reference standard)
Sigma-Aldrich
Tetramethylsilane, ≥99.0% (GC)
Supelco
Tetramethylsilane, analytical standard, suitable for NMR (spectroscopy), ACS reagent
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Tetramethylsilane, electronic grade, ≥99.99% trace metals basis
Sigma-Aldrich
Methanol, Laboratory Reagent, ≥99.6%
Supelco
Methanol, analytical standard
Supelco
Residual Solvent - Acetonitrile(solution in DMSO), Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Methanol, Absolute - Acetone free
Sigma-Aldrich
Methanol, ACS spectrophotometric grade, ≥99.9%
Sigma-Aldrich
Tetramethylsilane, NMR grade, ACS reagent, ≥99.9%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, suitable as ACS-grade LC reagent, ≥99.9%
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%
Supelco
Acetonitrile(Neat), Pharmaceutical Secondary Standard; Certified Reference Material