Merck
  • Parallel synthesis of cell-penetrating peptide conjugates of PMO toward exon skipping enhancement in Duchenne muscular dystrophy.
Parallel synthesis of cell-penetrating peptide conjugates of PMO toward exon skipping enhancement in Duchenne muscular dystrophy.
Nucleic acid therapeutics (2014-11-21)
Liz O'Donovan, Itaru Okamoto, Andrey A Arzumanov, Donna L Williams, Peter Deuss, Michael J Gait
ABSTRACT
We describe two new methods of parallel chemical synthesis of libraries of peptide conjugates of phosphorodiamidate morpholino oligonucleotide (PMO) cargoes on a scale suitable for cell screening prior to in vivo analysis for therapeutic development. The methods represent an extension of the SELection of PEPtide CONjugates (SELPEPCON) approach previously developed for parallel peptide-peptide nucleic acid (PNA) synthesis. However, these new methods allow for the utilization of commercial PMO as cargo with both C- and N-termini unfunctionalized. The synthetic methods involve conjugation in solution phase, followed by rapid purification via biotin-streptavidin immobilization and subsequent reductive release into solution, avoiding the need for painstaking high-performance liquid chromatography purifications. The synthesis methods were applied for screening of PMO conjugates of a 16-member library of variants of a 10-residue ApoE peptide, which was suggested for blood-brain barrier crossing. In this work the conjugate library was tested in an exon skipping assay using skeletal mouse mdx cells, a model of Duchene's muscular dystrophy where higher activity peptide-PMO conjugates were identified compared with the starting peptide-PMO. The results demonstrate the power of the parallel synthesis methods for increasing the speed of optimization of peptide sequences in conjugates of PMO for therapeutic screening.
Sigma-Aldrich
Ultrapure Acetonitrile
Dimethyl sulfoxide, European Pharmacopoeia (EP) Reference Standard
USP
Residual Solvent Class 2 - Acetonitrile, United States Pharmacopeia (USP) Reference Standard
USP
Methyl alcohol, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Acetonitrile, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Dimethyl sulfoxide, suitable for HPLC, ≥99.7%
Sigma-Aldrich
Trifluoroacetic acid, suitable for HPLC, ≥99.0%
Sigma-Aldrich
Acetonitrile, HPLC Plus, ≥99.9%
Sigma-Aldrich
Acetonitrile, suitable for HPLC, gradient grade, ≥99.9%
USP
Dimethyl sulfoxide, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Trifluoroacetic acid, ReagentPlus®, 99%
Sigma-Aldrich
Acetonitrile, ReagentPlus®, 99%
Sigma-Aldrich
Dimethyl sulfoxide, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Acetonitrile, biotech. grade, ≥99.93%
Sigma-Aldrich
Acetonitrile, ACS reagent, ≥99.5%
Sigma-Aldrich
Dimethyl sulfoxide, ACS reagent, ≥99.9%
Sigma-Aldrich
Trifluoroacetic acid, ≥99%, for protein sequencing
Sigma-Aldrich
Acetonitrile, suitable for DNA synthesis, ≥99.9% (GC)
Sigma-Aldrich
Trifluoroacetic acid, puriss. p.a., suitable for HPLC, ≥99.0% (GC)
Sigma-Aldrich
Dimethyl sulfoxide solution, 50 wt. % in H2O
Supelco
trans-Sinapic acid, analytical standard
Sigma-Aldrich
Phenylacetic acid, ≥99%, FCC, FG
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.6%, ReagentPlus®
Sigma-Aldrich
Methanol solution, suitable for NMR (reference standard), 4% in methanol-d4 (99.8 atom % D), NMR tube size 3 mm × 8 in.
Sigma-Aldrich
α-Cyano-4-hydroxycinnamic acid, 99%
Sigma-Aldrich
Dimethyl sulfoxide, anhydrous, ≥99.9%
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
Sigma-Aldrich
Phenylacetic acid, 99%
Supelco
α-Cyano-4-hydroxycinnamic acid, suitable for matrix substance for MALDI-MS, ≥99.0% (HPLC)
Sigma-Aldrich
8-Octanoyloxypyrene-1,3,6-trisulfonic acid trisodium salt, suitable for fluorescence, ≥90% (HPCE)