Merck
  • Identification of targets of Twist1 transcription factor in thyroid cancer cells.
Identification of targets of Twist1 transcription factor in thyroid cancer cells.
The Journal of clinical endocrinology and metabolism (2014-05-23)
Gennaro Di Maro, Francesca Maria Orlandella, Tammaro Claudio Bencivenga, Paolo Salerno, Clara Ugolini, Fulvio Basolo, Roberta Maestro, Giuliana Salvatore
ABSTRACT
Anaplastic thyroid carcinoma (ATC) is one of the most aggressive human tumors. Twist1 is a basic helix-loop-helix transcription factor involved in cancer development and progression. We showed that Twist1 affects thyroid cancer cell survival and motility. We aimed to identify Twist1 targets in thyroid cancer cells. Transcriptional targets of Twist1 were identified by gene expression profiling the TPC-Twist1 cells in comparison with control cells. Functional studies were performed by silencing in TPC-Twist1 and in CAL62 cells the top 10 upregulated genes and by evaluating cell proliferation, survival, migration, and invasion. Chromatin immunoprecipitation was performed to verify direct binding of Twist1 to target genes. Quantitative RT-PCR was applied to study the expression level of Twist1 target genes in human thyroid carcinoma samples. According to the gene expression profile, the top functions enriched in TPC-Twist1 cells were cellular movement, cellular growth and proliferation, and cell death and survival. Silencing of the top 10 upregulated genes reduced viability of TPC-Twist1 and of CAL62 cells. Silencing of COL1A1, KRT7, and PDZK1 also induced cell death. Silencing of HS6ST2, THRB, ID4, RHOB, and PDZK1IP also impaired migration and invasion of TPC-Twist1 and of CAL62 cells. Chromatin immunoprecipitation showed that Twist1 directly binds the promoter of the top 10 upregulated genes. Quantitative RT-PCR showed that HS6ST2, COL1A1, F2RL1, LEPREL1, PDZK1, and PDZK1IP1 are overexpressed in thyroid carcinoma samples compared with normal thyroids. We identified a set of genes that mediates Twist1 biological effects in thyroid cancer cells.
Sigma-Aldrich
Trypan Blue solution, 0.4%, liquid, sterile-filtered, suitable for cell culture
Sigma-Aldrich
Trypan Blue solution, 0.4%, suitable for microscopy
Sigma-Aldrich
MISSION® esiRNA, targeting human COL1A1
Sigma-Aldrich
MISSION® esiRNA, targeting mouse Col1a1
Sigma-Aldrich
Trypan Blue, ≥80% (HPLC), Dye content 60 %
Sigma-Aldrich
Trypan Blue, powder, BioReagent, suitable for cell culture