Merck
  • Dissecting the dynamic changes of 5-hydroxymethylcytosine in T-cell development and differentiation.
Dissecting the dynamic changes of 5-hydroxymethylcytosine in T-cell development and differentiation.
Proceedings of the National Academy of Sciences of the United States of America (2014-07-30)
Ageliki Tsagaratou, Tarmo Äijö, Chan-Wang J Lio, Xiaojing Yue, Yun Huang, Steven E Jacobsen, Harri Lähdesmäki, Anjana Rao
ABSTRACT
The discovery of Ten Eleven Translocation proteins, enzymes that oxidize 5-methylcytosine (5mC) in DNA, has revealed novel mechanisms for the regulation of DNA methylation. We have mapped 5-hydroxymethylcytosine (5hmC) at different stages of T-cell development in the thymus and T-cell differentiation in the periphery. We show that 5hmC is enriched in the gene body of highly expressed genes at all developmental stages and that its presence correlates positively with gene expression. Further emphasizing the connection with gene expression, we find that 5hmC is enriched in active thymus-specific enhancers and that genes encoding key transcriptional regulators display high intragenic 5hmC levels in precursor cells at those developmental stages where they exert a positive effect. Our data constitute a valuable resource that will facilitate detailed analysis of the role of 5hmC in T-cell development and differentiation.
Product Description
Supelco
Dehydrated Alcohol, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Ethanol solution, certified reference material, 2000 μg/mL in methanol
Sigma-Aldrich
3-Methyl-1-butanol, anhydrous, ≥99%
Sigma-Aldrich
3-Methyl-1-butanol, reagent grade, 98%
USP
Dehydrated Alcohol, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
3-Methyl-1-butanol, ACS reagent, ≥98.5%
Sigma-Aldrich
Ethyl alcohol, Pure 190 proof, for molecular biology
Supelco
3-Methyl-1-butanol, analytical standard
Sigma-Aldrich
Phenol, BioUltra, Molecular Biology, TE-saturated, ~73% (T)
Supelco
Phenol solution, certified reference material, 500 μg/mL in methanol
Supelco
Phenol solution, 5000 μg/mL in methanol, certified reference material
Sigma-Aldrich
Phenol, unstabilized, purified by redistillation, ≥99%
Sigma-Aldrich
Phenol solution, Equilibrated with 10 mM Tris HCl, pH 8.0, 1 mM EDTA, BioReagent, Molecular Biology
Sigma-Aldrich
Phenol solution, Saturated with 0.01 M citrate buffer, pH 4.3 ± 0.2, BioReagent, Molecular Biology
Sigma-Aldrich
Liquified Phenol, ≥89.0%
Sigma-Aldrich
Phenol, Molecular Biology
Sigma-Aldrich
Phenol, BioXtra, ≥99.5% (GC)
Sigma-Aldrich
3-Methylbutanol, BioReagent, ≥98.5%, Molecular Biology
Sigma-Aldrich
Isoamyl alcohol, ≥98%, FG
Sigma-Aldrich
Phenol, ≥99%
Sigma-Aldrich
Isoamyl alcohol, natural, ≥98%, FG
Sigma-Aldrich
Phenol, BioUltra, Molecular Biology, ≥99.5% (GC)
Supelco
Phenol solution, 100 μg/mL in acetonitrile, PESTANAL®, analytical standard
Sigma-Aldrich
Phenol, ≥96.0% (calc. on dry substance, T)
Sigma-Aldrich
Phenol, puriss. p.a., ACS reagent, reag. Ph. Eur., 99.0-100.5%
Sigma-Aldrich
Phenol, unstabilized, ReagentPlus®, ≥99%
Sigma-Aldrich
Phenol, ACS reagent, ≥99.0%
Supelco
3-Methylbutanol, analytical standard
Supelco
Ethanol, standard for GC
Sigma-Aldrich
Phenol, puriss., meets analytical specification of Ph. Eur., BP, USP, ≥99.5% (GC), crystalline (detached)