Merck
  • Treatment with amnion-derived cellular cytokine solution (ACCS) induces persistent motor improvement and ameliorates neuroinflammation in a rat model of penetrating ballistic-like brain injury.
Treatment with amnion-derived cellular cytokine solution (ACCS) induces persistent motor improvement and ameliorates neuroinflammation in a rat model of penetrating ballistic-like brain injury.
Restorative neurology and neuroscience (2015-01-16)
Ying Deng-Bryant, Ryan D Readnower, Lai Yee Leung, Tracy L Cunningham, Deborah A Shear, Frank C Tortella
ABSTRACT
The present work compared the behavioral outcomes of ACCS therapy delivered either intravenously (i.v.) or intracerebroventricularly (i.c.v.) after penetrating ballistic-like brain injury (PBBI). Histological markers for neuroinflammation and neurodegeneration were employed to investigate the potential therapeutic mechanism of ACCS. Experiment-1, ACCS was administered either i.v. or i.c.v. for 1 week post-PBBI. Outcome metrics included behavioral (rotarod and Morris water maze) and gross morphological assessments. Experiment-2, rats received ACCS i.c.v for either 1 or 2 weeks post-PBBI. The inflammatory response was determined by immunohistochemistry for neutrophils and microglia reactivity. Neurodegeneration was visualized using silver staining. Both i.v. and i.c.v. delivery of ACCS improved motor outcome but failed to improve cognitive outcome or tissue sparing. Importantly, only i.c.v. ACCS treatment produced persistent motor improvements at a later endpoint. The i.c.v. ACCS treatment significantly reduced PBBI-induced increase in myeloperoxidase (MPO) and ionized calcium binding adaptor molecule 1 (Iba1) expression. Concomitant reduction of both Iba1 and silver staining were detected in corpus callosum with i.c.v. ACCS treatment. ACCS, as a treatment for TBI, showed promise with regard to functional (motor) recovery and demonstrated strong capability to modulate neuroinflammatory responses that may underline functional recovery. However, the majority of beneficial effects appear restricted to the i.c.v. route of ACCS delivery, which warrants future studies examining delivery routes (e.g. intranasal delivery) which are more clinically viable for the treatment of TBI.
Sigma-Aldrich
Ethanol, purum, fine spirit, denaturated with 4.8% methanol, F25 METHYL1, ~96% (based on denaturant-free substance)
Sigma-Aldrich
Sodium thiosulfate, purum p.a., anhydrous, ≥98.0% (RT)
Sigma-Aldrich
o-Xylene, reagent grade, ≥98.0%
Sigma-Aldrich
Ethyl alcohol, Pure 200 proof, Molecular Biology
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, meets USP testing specifications
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, ACS reagent, ≥99.5%
Sigma-Aldrich
Sodium thiosulfate, ReagentPlus®, 99%
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, anhydrous, ≥99.5%
Sigma-Aldrich
Ethanol, absolute, sales not in Germany, ≥99.8% (vol.)
Sigma-Aldrich
Sodium thiosulfate, ≥99.99% trace metals basis
Sigma-Aldrich
Ethanol, tested according to Ph. Eur.
Supelco
Ethanol, standard for GC
Sigma-Aldrich
Ethanol
Supelco
o-Xylene, analytical standard
Sigma-Aldrich
o-Xylene, anhydrous, 97%
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, ACS spectrophotometric grade, 95.0%
Supelco
o-Xylene, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Ethanol, BioUltra, Molecular Biology, ≥99.8%, (absolute alcohol, without additive, A15 o1)
Sigma-Aldrich
Hydrogen peroxide solution, 30 % (w/w) in H2O, contains stabilizer
Supelco
Ethanol solution, certified reference material, 2000 μg/mL in methanol
Sigma-Aldrich
Hydrogen peroxide solution, tested according to Ph. Eur.
Millipore
Hydrogen peroxide solution, 3%, suitable for microbiology
Supelco
Hydrogen peroxide solution, ≥30%, for trace analysis
Sigma-Aldrich
Hydrogen peroxide solution, 34.5-36.5%
Sigma-Aldrich
Sodium Thiosulfate Solution, 2 g/dL in deionized water
Supelco
Dehydrated Alcohol, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Hydrogen peroxide solution, 30 % (w/w), for ultratrace analysis
USP
Dehydrated Alcohol, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Ethyl alcohol, Pure 190 proof, for molecular biology
Sigma-Aldrich
Hydrogen peroxide solution, contains inhibitor, 30 wt. % in H2O, meets USP testing specifications