- A modified method for the estimation of total carbonyl compounds in heated and frying oils using 2-propanol as a solvent.
A modified method for the estimation of total carbonyl compounds in heated and frying oils using 2-propanol as a solvent.
Journal of the American Oil Chemists' Society (2001)
Endo Y, et al.