- Significant enhancement of the Stille reaction with a new combination of reagents-copper(I) iodide with cesium fluoride.
Significant enhancement of the Stille reaction with a new combination of reagents-copper(I) iodide with cesium fluoride.
Chemistry (Weinheim an der Bergstrasse, Germany) (2005-03-24)
Simon P H Mee, Victor Lee, Jack E Baldwin
PMID15786503
ABSTRACT
The combination of copper(I) iodide and cesium fluoride significantly enhances the Stille reaction. After extensive optimisation, a variety of electronically unfavourable and sterically hindered substrates were coupled in very high yields under mild conditions.