Merck
Upstream binding factor inhibits herpes simplex virus replication.
Virology (2015-05-13)
Gabriel Ouellet Lavallée, Angela Pearson
ABSTRACT
Herpes simplex virus 1 (HSV-1) infection induces changes to the host cell nucleus including relocalization of the cellular protein Upstream Binding Factor (UBF) from the nucleolus to viral replication compartments (VRCs). Herein, we tested the hypothesis that UBF is recruited to VRCs to promote viral DNA replication. Surprisingly, infection of UBF-depleted HeLa cells with HSV-1 or HSV-2 produced higher viral titers compared to controls. Reduced expression of UBF also led to a progressive increase in the relative amount of HSV-1 DNA versus controls, and increased levels of HSV-1 ICP27 and TK mRNA and protein, regardless of whether viral DNA replication was inhibited or not. Our results suggest that UBF can inhibit gene expression from viral DNA prior to its replication. A similar but smaller effect on viral titers was observed in human foreskin fibroblasts. This is the first report of UBF having a restrictive effect on replication of a virus.
Sigma-Aldrich
Acrylamide, Molecular Biology, ≥99% (HPLC)
Sigma-Aldrich
MISSION® esiRNA, targeting human UBTF
Sigma-Aldrich
Acrylamide, suitable for electrophoresis, ≥99% (HPLC), powder
Sigma-Aldrich
Acrylamide, suitable for electrophoresis, ≥99%
Sigma-Aldrich
Anti-UBTF antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Sigma-Aldrich
Acrylamide solution, 40%, suitable for electrophoresis, sterile-filtered
Sigma-Aldrich
MISSION® esiRNA, targeting mouse Ubtf