Sigma-Aldrich
Manganese, chips, thickness <2.0 mm, 99%
Sigma-Aldrich
Manganese, powder, −325 mesh, ≥99% trace metals basis
Sigma-Aldrich
Selenium, pellets, <5 mm particle size, ≥99.999% trace metals basis
Sigma-Aldrich
Manganese, powder, ≥99.9% trace metals basis
Sigma-Aldrich
Selenium, powder, −100 mesh, ≥99.5% trace metals basis
Sigma-Aldrich
Selenium, pellets, <5 mm, ≥99.99% trace metals basis
Sigma-Aldrich
Selenium, powder, −100 mesh, 99.99% trace metals basis
Manganese, foil, not light tested, 25x25mm, thickness 0.008mm, permanent polyester support, 98.7%
Manganese, foil, not light tested, 50x50mm, thickness 0.02mm, permanent polyester support, 98.7%
Manganese, foil, not light tested, 25x25mm, thickness 0.05mm, permanent polyester support, 98.7%
Manganese, foil, not light tested, 50x50mm, thickness 0.001mm, permanent polyester support, 98.7%
Manganese, rod, 10mm, diameter 4.0mm, cast, 99.5%
Manganese, rod, 50mm, diameter 4.0mm, cast, 99.5%
Manganese, foil, not light tested, 50x50mm, thickness 0.015mm, permanent polyester support, 98.7%
Manganese, foil, not light tested, 50x50mm, thickness 0.002mm, permanent polyester support, 98.7%
Manganese, foil, not light tested, 25x25mm, thickness 0.007mm, permanent polyester support, 98.7%
Manganese, foil, not light tested, 25x25mm, thickness 0.025mm, permanent polyester support, 98.7%
Manganese, foil, not light tested, 50x50mm, thickness 0.01mm, permanent polyester support, 98.7%
Manganese, foil, not light tested, 25x25mm, thickness 0.004mm, permanent polyester support, 98.7%
Manganese, foil, not light tested, 50x50mm, thickness 0.05mm, permanent polyester support, 98.7%
Manganese, foil, not light tested, 50x50mm, thickness 0.004mm, permanent polyester support, 98.7%
Manganese, foil, not light tested, 50x50mm, thickness 0.007mm, permanent polyester support, 98.7%
Manganese, foil, not light tested, 50x50mm, thickness 0.0025mm, permanent polyester support, 98.7%
Manganese, foil, not light tested, 50x50mm, thickness 0.003mm, permanent polyester support, 98.7%
Manganese, foil, not light tested, 25x25mm, thickness 0.002mm, permanent polyester support, 98.7%
Manganese, foil, not light tested, 25x25mm, thickness 0.0025mm, permanent polyester support, 98.7%
Manganese, rod, 50mm, diameter 6.35mm, cast, 99.5%
Manganese, foil, not light tested, 25x25mm, thickness 0.005mm, permanent polyester support, 98.7%
Selenium, pellets, < 5mm, ≥99.999%
Manganese, foil, not light tested, 25x25mm, thickness 0.009mm, permanent polyester support, 98.7%