Merck
  • Elevated Expression of the Delta-Subunit of Epithelial Sodium Channel in Temporal Lobe Epilepsy Patients and Rat Model.
Elevated Expression of the Delta-Subunit of Epithelial Sodium Channel in Temporal Lobe Epilepsy Patients and Rat Model.
Journal of molecular neuroscience : MN (2015-08-02)
Zhonggui Li, Xiujuan Mi, Yan Xiong, Xin Xu, Xuefeng Wang, Liang Wang
ABSTRACT
Epilepsy is one of the most common, chronic, neurological diseases. The pathology of epilepsy is based on abnormal synchronization of neuronal discharges. Epithelial sodium channels, which are constitutively active, non-voltage-gated, highly selective sodium channels belonging to the epithelial sodium channel/degenerin (ENaC/deg) family, contribute to resting membrane potential modulation and subsequent neuronal excitability by providing a sodium influx pathway. Different from the other three subunits, δ ENaC expression is prominent in the human brain cortex and restricted to neurons. The aim of this study was to investigate the expression pattern of δ ENaC in patients with temporal lobe epilepsy (TLE) and in a pilocarpine-induced rat model of epilepsy. Adopting immunohistochemistry, immunofluorescence, and western blot analysis, we found that δ ENaC was restricted to neurons in the temporal neocortices of TLE patients and the cortices and hippocampus of pilocarpine-induced epilepsy rats, which were similar to the corresponding controls. However, δ ENaC expression was significantly elevated in TLE patients and the pilocarpine-induced epileptic rats. The physiological role, the unchanged localization, and the elevated expression of δ ENaC suggested it could play an important role in epilepsy.
Sigma-Aldrich
DAPI, for nucleic acid staining
Sigma-Aldrich
Lamotrigine, ≥98%, powder
Sigma-Aldrich
Lithium-7Li chloride, 99 atom % 7Li, 99% (CP)
Sigma-Aldrich
Lithium chloride, Molecular Biology, ≥99%
Sigma-Aldrich
Pyrrolidone hydrotribromide, 97%
Sigma-Aldrich
Lithium chloride, powder, ≥99.98% trace metals basis
Sigma-Aldrich
Lithium chloride, AnhydroBeads, −10 mesh, ≥99.9% trace metals basis
Sigma-Aldrich
Lithium chloride, BioUltra, Molecular Biology, anhydrous, ≥99.0% (AT)
Sigma-Aldrich
Lithium chloride, AnhydroBeads, −10 mesh, 99.998% trace metals basis
Sigma-Aldrich
Lithium chloride, anhydrous, free-flowing, Redi-Dri, ACS reagent, ≥99%
Sigma-Aldrich
Lithium chloride, ACS reagent, ≥99%
Sigma-Aldrich
Carbamazepine, powder
Sigma-Aldrich
Carbamazepine, meets USP testing specifications
Sigma-Aldrich
Lithium chloride, anhydrous, free-flowing, Redi-Dri, ReagentPlus®, 99%
Sigma-Aldrich
Lithium chloride, BioXtra, ≥99.0% (titration)
Sigma-Aldrich
Lithium chloride solution, 8 M, Molecular Biology, ≥99%
Sigma-Aldrich
Lithium chloride, ReagentPlus®, 99%