Merck
  • Physical Stimuli-Induced Chondrogenic Differentiation of Mesenchymal Stem Cells Using Magnetic Nanoparticles.
Physical Stimuli-Induced Chondrogenic Differentiation of Mesenchymal Stem Cells Using Magnetic Nanoparticles.
Advanced healthcare materials (2015-04-08)
Boram Son, Hwan D Kim, Minsoo Kim, Jeong Ah Kim, Jinkyu Lee, Heungsoo Shin, Nathaniel S Hwang, Tai Hyun Park
ABSTRACT
Chondrogenic commitments of mesenchymal stem cells (MSCs) require 3D cellular organization. Furthermore, recent progresses in bioreactor technology have contributed to the development of various biophysical stimulation platforms for efficient cartilage tissue formation. Here, an approach is reported to drive 3D cellular organization and enhance chondrogenic commitment of bone-marrow-derived human mesenchymal stem cells (BM-hMSCs) via magnetic nanoparticle (MNP)-mediated physical stimuli. MNPs isolated from Magnetospirillum sp. AMB-1 are endocytosed by the BM-hMSCs in a highly efficient manner. MNPs-incorporated BM-hMSCs are pelleted and then subjected to static magnetic field and/or magnet-derived shear stress. Magnetic-based stimuli enhance level of sulfated glycosaminoglycan (sGAG) and collagen synthesis, and facilitate the chondrogenic differentiation of BM-hMSCs. In addition, both static magnetic field and magnet-derived shear stress applied for the chondrogenic differentiation of BM-hMSCs do not show increament of hypertrophic differentiation. This MNP-mediated physical stimulation platform demonstrates a promising strategy for efficient cartilage tissue engineering.
Sigma-Aldrich
Selenous acid, 99.999% trace metals basis
Sigma-Aldrich
Dexamethasone, ≥98% (HPLC), powder
Sigma-Aldrich
Dexamethasone, meets USP testing specifications
Sigma-Aldrich
Dexamethasone, powder, BioReagent, suitable for cell culture, ≥97%
Sigma-Aldrich
Dexamethasone, powder, γ-irradiated, BioXtra, suitable for cell culture, ≥80% (HPLC)
Sigma-Aldrich
Selenous acid, 98%
SAFC
L-Proline
Sigma-Aldrich
Sodium pyruvate, powder, BioXtra, suitable for mouse embryo cell culture
Sigma-Aldrich
Sodium pyruvate, Hybri-Max, powder, suitable for hybridoma
Sigma-Aldrich
Sodium pyruvate, ReagentPlus®, ≥99%
Sigma-Aldrich
Sodium pyruvate, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥99%
Sigma-Aldrich
L-Proline, ReagentPlus®, ≥99% (HPLC)
Sigma-Aldrich
L-Proline, from non-animal source, meets EP, USP testing specifications, suitable for cell culture
Sigma-Aldrich
Sodium pyruvate, BioXtra, ≥99%
Sigma-Aldrich
L-Proline, 99%, FCC, FG
Sigma-Aldrich
L-Proline, BioUltra, ≥99.5% (NT)
Sigma-Aldrich
Sodium pyruvate, anhydrous, free-flowing, Redi-Dri, ReagentPlus®, ≥99%