Merck
  • Effectiveness of levulinic acid and sodium dodecyl sulfate employed as a sanitizer during harvest or packing of cantaloupes contaminated with Salmonella Poona.
Effectiveness of levulinic acid and sodium dodecyl sulfate employed as a sanitizer during harvest or packing of cantaloupes contaminated with Salmonella Poona.
International journal of food microbiology (2015-05-23)
Cathy C Webb, Marilyn C Erickson, Lindsey E Davey, Michael P Doyle
ABSTRACT
Freshly harvested Eastern variety cantaloupes (Cucumis melo L. var. reticulatus cv. Athena) were subjected to three different harvest and wash treatments to examine conditions under which the efficacy of the sanitizer, levulinic acid (LV) plus sodium dodecyl sulfate (SDS), could be enhanced to reduce Salmonella contamination. In treatment set one, cantaloupes were spot inoculated with Salmonella enterica serovar Poona (prepared from solid or liquid media cultures) before or after a 1-min dip treatment in LV (2.5, 5.0, 7.5, or 10%) and 2.5% SDS. S. Poona initial populations on rind tissue (4.26-5.04 log CFU/sample) were reduced to detection by enrichment culture when cantaloupes were subsequently exposed to any of the LV/SDS solutions. When S. Poona was introduced after cantaloupes had been dip-treated, greater decreases in pathogen populations at the stem scar were observed when cantaloupes were treated with increasing concentrations of LV. In treatment set two, the response of S. Poona dip-treated with 5% LV/2.5% SDS was compared to a simulated commercial dump tank treatment incorporating 200 ppm chlorine as well as a two-stage treatment employing both the chlorine tank and LV/SDS dip treatments. S. Poona levels (log CFU/sample or # positive by enrichment culture/# analyzed) after treatments were 5.25, 3.07, 7/10, 5/10 (stem scar) and 3.90, 25/40, 28/40, 20/40 (rind) for non-treated, chlorine tank, LV/SDS dip, and tank plus dip treatments, respectively. In treatment set three, freshly harvested cantaloupes were first treated in the field using a needle-free stem scar injection (200 μl, 7.5% LV/1.0% SDS, 60 psi) and a cantaloupe spray (30 ml, 7.5% LV/0.5% SDS). Cantaloupe stem scar and rind tissue were then spot-inoculated with S. Poona using either a liquid or soil-based medium followed by a simulated dump tank treatment incorporating either 200 ppm chlorine or 5% LV/2% SDS. S. Poona inoculated on field-treated cantaloupe rind decreased by 4.7 and 5.31 (liquid) and 3.27 and 3.36 (soil) log CFU/sample after simulated chlorine and LV/SDS tank treatments, respectively. In the case of stem scar tissue, S. Poona populations exhibited a 1.0 log greater reduction when cantaloupes were treated with LV/SDS compared to chlorine in the dump tank (P<0.05). Based on this study, application of multiple hurdles is warranted, as additional decreases in S. Poona populations were obtained when cantaloupes were subjected to a chlorine dump tank followed by a LV/SDS dip treatment.
Sigma-Aldrich
Sodium thiosulfate, ≥99.99% trace metals basis
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, Molecular Biology, 10% in H2O
Sigma-Aldrich
Ampicillin, anhydrous, 96.0-102.0% (anhydrous basis)
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, 20% in H2O
Sigma-Aldrich
Chlorine, ≥99.5%
Sigma-Aldrich
Levulinic acid, 98%
Sigma-Aldrich
Sodium dodecyl sulfate, ≥90% ((Assay))
Sigma-Aldrich
Sodium dodecyl sulfate, ≥98.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, BioUltra, Molecular Biology, ≥99.0% (GC)
Sigma-Aldrich
Sulfuric acid, 99.999%
Sigma-Aldrich
Levulinic acid, ≥97%, FG
Sigma-Aldrich
Sodium dodecyl sulfate, tested according to NF, mixture of sodium alkyl sulfates consisting mainly of sodium dodecyl sulfate
Sigma-Aldrich
Sodium dodecyl sulfate, ACS reagent, ≥99.0%
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, Molecular Biology, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ReagentPlus®, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, 92.5-100.5% based on total alkyl sulfate content basis
Sigma-Aldrich
Sodium dodecyl sulfate, BioXtra, ≥99.0% (GC)
Sigma-Aldrich
Sodium Thiosulfate Solution, 2 g/dL in deionized water
Sigma-Aldrich
Ampicillin, meets USP testing specifications
Sigma-Aldrich
Sodium pyruvate, ReagentPlus®, ≥99%
Sigma-Aldrich
Sodium pyruvate, Hybri-Max, powder, suitable for hybridoma
Supelco
Sodium dodecyl sulfate, dust-free pellets, suitable for electrophoresis, Molecular Biology, ≥99.0% (GC)
Sigma-Aldrich
Levulinic acid, natural, 99%, FG
Sigma-Aldrich
Sodium pyruvate, powder, BioXtra, suitable for mouse embryo cell culture
Sigma-Aldrich
Sodium pyruvate, BioXtra, ≥99%
Sigma-Aldrich
Sodium pyruvate, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥99%
Sigma-Aldrich
Sodium dodecyl sulfate, ≥99.0% (GC), dust-free pellets
Sigma-Aldrich
Sodium pyruvate, anhydrous, free-flowing, Redi-Dri, ReagentPlus®, ≥99%
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, Molecular Biology, ≥98.5% (GC), free-flowing, Redi-Dri