Merck
  • Using quantitative structure activity relationship models to predict an appropriate solvent system from a common solvent system family for countercurrent chromatography separation.
Using quantitative structure activity relationship models to predict an appropriate solvent system from a common solvent system family for countercurrent chromatography separation.
Journal of chromatography. A (2015-05-02)
Siân Marsden-Jones, Nicola Colclough, Ian Garrard, Neil Sumner, Svetlana Ignatova
ABSTRACT
Countercurrent chromatography (CCC) is a form of liquid-liquid chromatography. It works by running one immiscible solvent (mobile phase) over another solvent (stationary phase) being held in a CCC column using centrifugal force. The concentration of compound in each phase is characterised by the partition coefficient (Kd), which is the concentration in the stationary phase divided by the concentration in the mobile phase. When Kd is between approximately 0.2 and 2, it is most likely that optimal separation will be achieved. Having the Kd in this range allows the compound enough time in the column to be separated without resulting in a broad peak and long run time. In this paper we report the development of quantitative structure activity relationship (QSAR) models to predict logKd. The QSAR models use only the molecule's 2D structure to predict the molecular property logKd.
Sigma-Aldrich
Acetonitrile, electronic grade, 99.999% trace metals basis
Sigma-Aldrich
Ethyl acetate
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, anhydrous, ≥99.5%
Sigma-Aldrich
Ethyl acetate, ≥99%, FCC, FG
Sigma-Aldrich
Ethyl acetate, anhydrous, 99.8%
Sigma-Aldrich
Ethyl acetate, natural, ≥99%, FCC, FG
Sigma-Aldrich
Ethyl acetate, ReagentPlus®, ≥99.8%
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, ACS spectrophotometric grade, 95.0%
Sigma-Aldrich
Methanol solution, suitable for NMR (reference standard), 4% in methanol-d4 (99.8 atom % D), NMR tube size 3 mm × 8 in.
Sigma-Aldrich
Methanol, suitable for NMR (reference standard)
Sigma-Aldrich
Heptane, anhydrous, 99%
Sigma-Aldrich
Ammonia, anhydrous, ≥99.98%
Sigma-Aldrich
Ammonia solution, 0.4 M in THF
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Ammonia-14N, 99.99 atom % 14N
Sigma-Aldrich
Ammonia solution, 4 M in methanol