Merck
  • Study the effects of PLGA-PEG encapsulated amphotericin B nanoparticle drug delivery system against Leishmania donovani.
Study the effects of PLGA-PEG encapsulated amphotericin B nanoparticle drug delivery system against Leishmania donovani.
Drug delivery (2014-03-08)
Rishikesh Kumar, Ganesh Chandra Sahoo, Krishna Pandey, Vnr Das, Pradeep Das
ABSTRACT
Drug delivery systems are a promising technology to increase poor solubility and bioavailability of compounds. Therefore we have developed PLGA-PEG encapsulated amphotericin B nanoparticles (NPs) drug delivery technology to increase the solubility of amphotericin B and target the macrophage of infected tissues during visceral leishmaniasis. The structural characterization by transmission electron microscopy and dynamic light scattering revealed the nano-size of the particle (30-35 nanometers). Fourier transform infrared spectroscopy confirmed the PLGA-PEG encapsulation. The mean cytotoxic assay (0.0803 + 0.0253) of extracellular promastigote of PLGA-PEG encapsulated amphotericin B is significantly lower than that of amphotericin B (0.1134 + 0.0153) and inhibition of amastigotes in the splenic tissue was significantly more than with conventional amphotericin B (93.02 + 6.63 versus 74.42 +14.78). Amphotericin B encapsulated PLGA-PEG nanoparticles were found to be more effective than free amphotericin B in terms of therapeutic efficacy during in vitro and in vivo study.
Sigma-Aldrich
Dichloromethane, anhydrous, ≥99.8%, contains 40-150 ppm amylene as stabilizer
Sigma-Aldrich
N-(3-Dimethylaminopropyl)-N′-ethylcarbodiimide, ≥97.0% (T)
Sigma-Aldrich
1,2-Dichloroethane, anhydrous, 99.8%
Sigma-Aldrich
Dichloromethane, suitable for HPLC, ≥99.9%, contains 40-150 ppm amylene as stabilizer