Merck
  • Influence of water temperature and waterborne cadmium toxicity on growth performance and metallothionein-cadmium distribution in different organs of Nile tilapia, Oreochromis niloticus (L.).
Influence of water temperature and waterborne cadmium toxicity on growth performance and metallothionein-cadmium distribution in different organs of Nile tilapia, Oreochromis niloticus (L.).
Journal of thermal biology (2014-12-02)
Mohsen Abdel-Tawwab, Mohammed Wafeek
ABSTRACT
Cadmium (Cd) is believed to be one of the most abundant and ubiquitously distributed toxins in the aquatic system. This metal is released to the aquatic environment from both anthropogenic sources, such as industrial, agricultural and urban effluents as well as natural sources, such as rocks and soils. Otherwise, the temperature increase of water bodies, which has been observed due to global climatic changes, has been shown to increase Cd toxicity for several aquatic animal species including fish. In the present study, Nile tilapia, Oreochromis niloticus (L.), (26.0 ± 0.38 g) were reared at 20, 24, 28, or 32 °C and exposed to 0.0 or 0.5mg Cd/L for 8 weeks to investigate effects of water temperature, Cd toxicity and their interaction on fish performance as well as metallothionein (MT) and Cd distribution in different fish organs. It was found that fish reared in Cd-free group at 28 °C showed the optimum growth and feed intake, while Cd-exposed fish showed low growth and feed intake irrespective to water temperature. A synergetic relationship between water temperature and Cd toxicity was observed where Cd toxicity increased as water temperature increased and the worse growth was obtained in Cd-exposed fish reared at 32 °C. Additionally, the highest Cd residues in different fish organs were detected in Cd-exposed fish reared at 32 °C. Similarly, MT concentrations in different fish organs increased as water temperature increased especially in Cd-exposed fish groups. A high positive correlation between MT and Cd concentrations in fish organs was detected. The distribution of MT and Cd levels was in the order of liver>kidney>gills>muscles. The present study revealed that the optimum water temperature suitable for Nile tilapia growth is 28 °C. Additionally, Cd exposure had a deteriorate effect on the growth and health of Nile tilapia. This hazardous effect increased as water temperature increased. Further, liver and kidney were the prime sites of Cd accumulation, while Cd load in the muscles was the lowest as compared to the other investigated organs.
Sigma-Aldrich
Ammonia-14N, 99.99 atom % 14N
Sigma-Aldrich
Ammonia, anhydrous, ≥99.98%
Sigma-Aldrich
Ammonia solution, 4 M in methanol
Sigma-Aldrich
Ammonia solution, 0.4 M in THF