Merck
  • Elevations in T-helper-2-initiating cytokines (interleukin-33, interleukin-25 and thymic stromal lymphopoietin) in lesional skin from chronic spontaneous ('idiopathic') urticaria.
Elevations in T-helper-2-initiating cytokines (interleukin-33, interleukin-25 and thymic stromal lymphopoietin) in lesional skin from chronic spontaneous ('idiopathic') urticaria.
The British journal of dermatology (2014-12-20)
A B Kay, P Clark, M Maurer, S Ying
ABSTRACT
The mechanism of wealing in chronic spontaneous urticaria (CSU) is largely unknown. We previously demonstrated increased expression of T-helper 2 [interleukin (IL)-4 and IL-5] cytokines in skin biopsies from CSU. This suggested that Th2-initiating cytokines [IL-33, IL-25 and thymic stromal lymphopoietin (TSLP)], released through innate immune mechanisms, may play a role in pathogenesis. To identify Th2-initiating cytokines in lesional and nonlesional skin from patients with CSU and to compare the results with a control group. Paired biopsies (one from a 4-8 h spontaneous weal and one from uninvolved skin) were taken from eight patients with CSU and nine control subjects, and studied by immunohistochemistry and confocal microscopy. There were increases in IL-4(+) and IL-5(+) cells in lesional skin vs. controls (P = 0·03 and P < 0·001, respectively) and marked elevations in the numbers of IL-33(+), IL-25(+) and TSLP(+) cells in the dermis of lesional skin vs. both nonlesional skin (P = 0·002, P = 0·01 and P = 0·04, respectively) and controls (P = 0·001, P < 0·001 and P = 0·005, respectively). There was also a correlation between the numbers of IL-33(+) and IL-25(+) cells (r = 0·808, P = 0·015). IL-33 localized to CD31(+) endothelial cells, CD90(+) fibroblasts, CD68(+) macrophages and tryptase(+) mast cells, whereas IL-25 was expressed by epithelial cells, mast cells and major basic protein-positive eosinophils. IL-33 and IL-25 were constitutively expressed in the epidermis of both controls and patients with CSU. Increased expression of Th2-initiating cytokines in lesional skin in CSU suggests that innate pathways might play a role in the mechanism of wealing. As Th2-initiating cytokines play a role in mast cell activation, inflammation and vascular leakage in CSU, these findings may also have therapeutic implications.
Sigma-Aldrich
Hydrogen peroxide solution, 34.5-36.5%
Sigma-Aldrich
Hydrogen peroxide solution, 30 % (w/w) in H2O, contains stabilizer
Sigma-Aldrich
Fluorescein isothiocyanate isomer I, ≥97.5% (HPLC)
Sigma-Aldrich
IL-33 human, Animal-component free, recombinant, expressed in E. coli, ≥98% (SDS-PAGE), ≥98% (HPLC)
Sigma-Aldrich
3,3′-Diaminobenzidine, 97%
Sigma-Aldrich
Fluorescein 5(6)-isothiocyanate, BioReagent, suitable for fluorescence, mixture of 2 components, ≥90% (HPLC)
Sigma-Aldrich
Fluorescein 5(6)-isothiocyanate, ≥90% (HPLC)
Sigma-Aldrich
DAPI, for nucleic acid staining
Sigma-Aldrich
3,3′-Diaminobenzidine, 97% (HPLC)