Merck
  • Comparative analysis of type 2 diabetes-associated SNP alleles identifies allele-specific DNA-binding proteins for the KCNQ1 locus.
Comparative analysis of type 2 diabetes-associated SNP alleles identifies allele-specific DNA-binding proteins for the KCNQ1 locus.
International journal of molecular medicine (2015-05-09)
Masaki Hiramoto, Haruhide Udagawa, Atsushi Watanabe, Keisuke Miyazawa, Naoko Ishibashi, Miho Kawaguchi, Takashi Uebanso, Wataru Nishimura, Takao Nammo, Kazuki Yasuda
ABSTRACT
Although recent genome-wide association studies (GWAS) have been extremely successful, it remains a big challenge to functionally annotate disease‑associated single nucleotide polymorphisms (SNPs), as the majority of these SNPs are located in non‑coding regions of the genome. In this study, we described a novel strategy for identifying the proteins that bind to the SNP‑containing locus in an allele‑specific manner and successfully applied this method to SNPs in the type 2 diabetes mellitus susceptibility gene, potassium voltage‑gated channel, KQT‑like subfamily Q, member 1 (KCNQ1). DNA fragments encompassing SNPs, and risk or non‑risk alleles were immobilized onto the novel nanobeads and DNA‑binding proteins were purified from the nuclear extracts of pancreatic β cells using these DNA‑immobilized nanobeads. Comparative analysis of the allele-specific DNA-binding proteins indicated that the affinities of several proteins for the examined SNPs differed between the alleles. Nuclear transcription factor Y (NF‑Y) specifically bound the non‑risk allele of the SNP rs2074196 region and stimulated the transcriptional activity of an artificial promoter containing SNP rs2074196 in an allele‑specific manner. These results suggest that SNP rs2074196 modulates the affinity of the locus for NF‑Y and possibly induces subsequent changes in gene expression. The findings of this study indicate that our comparative method using novel nanobeads is effective for the identification of allele‑specific DNA‑binding proteins, which may provide important clues for the functional impact of disease‑associated non‑coding SNPs.
Sigma-Aldrich
Magnesium chloride solution, BioUltra, Molecular Biology, ~0.025 M in H2O
Sigma-Aldrich
Thymidine, ≥99.0% (HPLC)
Sigma-Aldrich
Magnesium chloride solution, BioUltra, Molecular Biology, ~1 M in H2O
Sigma-Aldrich
Glycerol solution, 83.5-89.5% (T)
Sigma-Aldrich
Thymidine, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Magnesium chloride solution, PCR Reagent, 25 mM MgCI2 solution for PCR
Sigma-Aldrich
Magnesium chloride solution, Molecular Biology, 1.00 M±0.01 M
Sigma-Aldrich
Thymidine, ≥99%
Sigma-Aldrich
Magnesium chloride solution, 0.1 M
Supelco
DL-Dithiothreitol solution, 1 M in H2O
Sigma-Aldrich
DL-Dithiothreitol solution, BioUltra, Molecular Biology, ~1 M in H2O
Sigma-Aldrich
Magnesium chloride solution, BioUltra, Molecular Biology, 2 M in H2O
Sigma-Aldrich
Magnesium chloride, AnhydroBeads, −10 mesh, 99.9% trace metals basis
Sigma-Aldrich
Magnesium chloride, AnhydroBeads, −10 mesh, 99.99% trace metals basis
Sigma-Aldrich
Glycerol, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for electrophoresis, ≥99% (GC)
Sigma-Aldrich
Glycerin, meets USP testing specifications
Sigma-Aldrich
Magnesium chloride, suitable for insect cell culture, BioReagent, ≥97.0%
Sigma-Aldrich
Magnesium chloride, ≥98%
Sigma-Aldrich
Iodoacetamide, ≥99% (NMR), crystalline
Sigma-Aldrich
Glycerol, ≥99.5%
Sigma-Aldrich
Glycerol, Molecular Biology, ≥99.0%
Sigma-Aldrich
Glycerol, BioXtra, ≥99% (GC)
Sigma-Aldrich
Magnesium chloride, powder, <200 μm
Sigma-Aldrich
Iodoacetamide, BioUltra
Sigma-Aldrich
Glycerol, BioUltra, Molecular Biology, anhydrous, ≥99.5% (GC)
Sigma-Aldrich
Iodoacetamide, Single use vial of 56 mg
Sigma-Aldrich
Glycerol, FCC, FG
SAFC
Iodoacetamide