Merck
  • Morphometric and neurochemical alterations found in l-BMAA treated rats.
Morphometric and neurochemical alterations found in l-BMAA treated rats.
Environmental toxicology and pharmacology (2015-05-24)
Estefanía de Munck, Emma Muñoz-Sáez, Begoña G Miguel, M Teresa Solas, Ana Martínez, Rosa M Arahuetes
ABSTRACT
Amyotrophic lateral sclerosis (ALS) is a neurodegenerative disease characterized by progressive muscle paralysis that reflects the motoneurons' degeneration. Several studies support the relationship between β-N-methylamino-l-alanine (l-BMAA), a neurotoxic amino acid produced by cyanobacteria and diatoms, and the sporadic occurrence of ALS and other neurodegenerative diseases. Therefore, the study of its neurotoxicity mechanisms has assumed great relevance in recent years. Recently, our research team has proposed a sporadic ALS animal model by l-BMAA administration in rats, which displays many pathophysiological features of human ALS. In this paper, we deepen the characterization of this model corroborating the occurrence of alterations present in ALS patients such as decreased muscle volume, thinning of the motor cortex, enlarged brain's lateral ventricles, and alteration of both bulbar nuclei and neurotransmitters' levels. Therefore, we conclude that l-BMAA treated rats could be a good model which mimics degenerative features that ALS causes in humans.
Sigma-Aldrich
Hydrogen chloride solution, 3 M in cyclopentyl methyl ether (CPME)
Sigma-Aldrich
Phthaldialdehyde, suitable for fluorescence, ≥99.0% (HPLC)
Sigma-Aldrich
3-Mercaptopropionic acid, ≥99.0% (HPLC)
Sigma-Aldrich
3-Mercaptopropionic acid, ≥99%
Sigma-Aldrich
Hydrochloric acid solution, for amino acid analysis, ~6 M in H2O
Sigma-Aldrich
Hydrochloric acid solution, 1.0 N, BioReagent, suitable for cell culture
Sigma-Aldrich
Hydrochloric acid, 36.5-38.0%, BioReagent, Molecular Biology
Sigma-Aldrich
Hydrochloric acid solution, 32 wt. % in H2O, FCC
Sigma-Aldrich
Phthaldialdehyde, ≥97% (HPLC), powder or crystals
Sigma-Aldrich
γ-Aminobutyric acid, ≥99%
Sigma-Aldrich
Xylazine, ≥99%
Sigma-Aldrich
Phthaldialdehyde, suitable for HPLC fluorimetric detection of amino acids, ≥99% (HPLC), powder or crystals
Sigma-Aldrich
Octylphosphonic acid, 97%
Supelco
Hydrochloric acid solution, volumetric, 0.1 M HCl (0.1N), endotoxin free
Sigma-Aldrich
Taurine, ≥98%, FG
SAFC
Taurine
Sigma-Aldrich
Taurine, suitable for cell culture, meets USP testing specifications
Sigma-Aldrich
Taurine, BioUltra, ≥99.5% (T)
Sigma-Aldrich
Taurine, ≥99%
Sigma-Aldrich
Hydrogen chloride, ReagentPlus®, ≥99%