Merck
  • Heterojunction synergies in titania-supported gold photocatalysts: implications for solar hydrogen production.
Heterojunction synergies in titania-supported gold photocatalysts: implications for solar hydrogen production.
ChemSusChem (2015-06-25)
Vedran Jovic, Kevin E Smith, Hicham Idriss, Geoffrey I N Waterhouse
ABSTRACT
The mixed-phase nature of P25 TiO2 (85 % anatase/15 % rutile) plays a key role in the high H2 production rates shown by Au/P25 TiO2 photocatalysts in alcohol/water systems. However, a full understanding of the synergistic charge transfer mechanisms between the TiO2 polymorphs that drive the high rates is yet to be realised. Here, we deconstruct P25 TiO2 into its component phases, functionalise the phases with Au nanoparticles and explore charge transfer in Au/TiO2 systems using EPR spectroscopy. EPR spectroscopy and photocatalytic data provide direct evidence that electrons excited across the rutile band gap move to anatase lattice traps through interfacial surface sites, which decreases electron-hole pair recombination and increases charge carrier availability for photoreactions. In particular, three-phase interfacial sites between Au, anatase and rutile appear to be H2 evolution "hot spots". The results isolate the origin of high photocatalytic H2 production rates seen in Au/P25 TiO2 systems.
Sigma-Aldrich
Ammonia, anhydrous, ≥99.98%
Sigma-Aldrich
Ammonia solution, 4 M in methanol
Sigma-Aldrich
Ammonia solution, 0.4 M in THF
Sigma-Aldrich
Urea-12C, 99.9 atom % 12C
Sigma-Aldrich
Urea solution, 40 % (w/v) in H2O
Sigma-Aldrich
Hydrogen peroxide solution, 30 % (w/w) in H2O, contains stabilizer
Sigma-Aldrich
Hydrogen peroxide solution, 34.5-36.5%
Sigma-Aldrich
Urea solution, BioUltra, ~8 M in H2O
Sigma-Aldrich
Ammonia-14N, 99.99 atom % 14N
Supelco
Urea, 8 M (after reconstitution with 16 mL high purity water)
Sigma-Aldrich
Urea, ReagentPlus®, ≥99.5%, pellets
Sigma-Aldrich
Urea, BioXtra, pH 7.5-9.5 (20 °C, 5 M in H2O)
Sigma-Aldrich
Urea, meets USP testing specifications
Sigma-Aldrich
Urea, suitable for electrophoresis
Sigma-Aldrich
Urea, ACS reagent, 99.0-100.5%
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, anhydrous, ≥99.5%
Sigma-Aldrich
Urea, puriss. p.a., ACS reagent, reag. Ph. Eur., ≥99%
Sigma-Aldrich
Urea, puriss., meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%, 99.0-101.0% (calc. on dry substance)
Sigma-Aldrich
Ethanol, absolute, sales not in Germany, ≥99.8% (vol.)
Sigma-Aldrich
Urea, BioUltra, Molecular Biology, 99% (T)
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, ACS spectrophotometric grade, 95.0%
Sigma-Aldrich
Urea, powder, BioReagent, Molecular Biology, suitable for cell culture