Merck
  • Neurotrophine-3 may contribute to neuronal differentiation of mesenchymal stem cells through the activation of the bone morphogenetic protein pathway.
Neurotrophine-3 may contribute to neuronal differentiation of mesenchymal stem cells through the activation of the bone morphogenetic protein pathway.
Cellular & molecular biology letters (2015-07-26)
Lei Li, Yunpeng Li, Hongkun Jiang
ABSTRACT
We investigated whether neurotrophin-3 (NT-3) can promote differentiation of mouse bone mesenchymal stem cells (MSCs) into neurons via the bone morphogenetic protein pathway. MSCs were prepared from rat bone marrow and either transfected with pIRES2-EGFP or pIRES2-EGFP-NT-3 or treated with bone morphogenetic protein 4. The pIRES2-EGFP-NT-3-transfected MSCs further underwent noggin treatment or siRNA-mediated knockout of the TrkC gene or were left untreated. Immunofluorescence staining, real-time PCR and Western blot analyses were performed to evaluate the transcription and expression of neural-specific genes and BMP-Smad signaling. MSCs were efficiently transduced by the NT-3 gene via pIRES2-EGFP vectors. pIRES2- EGFP-NT-3 could initiate the transcription and expression of neural-specific genes, including nestin, NSE and MAP-2, and stimulate BMP-Smad signaling. The transcription and expression of neural-specific genes and BMP-Smad signaling were significantly suppressed by siRNA-mediated knockdown of the TrkC gene of MSCs. These findings suggest that the BMP signaling pathway may be a key regulatory point in NT-3-transfected neuronal differentiation of MSCs. The BMP and neurotrophin pathways contribute to a tightly regulated signaling network that directs the precise connections between neuronal differentiation of MSCs and their targets.
Sigma-Aldrich
L-Ascorbic acid, FCC, FG
Sigma-Aldrich
L-Ascorbic acid, ACS reagent, ≥99%
Sigma-Aldrich
1,5,7-Triazabicyclo[4.4.0]dec-5-ene, 98%
Sigma-Aldrich
MISSION® esiRNA, targeting human NTRK3 (1)
Sigma-Aldrich
L-Ascorbic acid, puriss. p.a., ≥99.0% (RT)
Sigma-Aldrich
L-Ascorbic acid, 99%
Sigma-Aldrich
L-Ascorbic acid, puriss. p.a., ACS reagent, reag. ISO, Ph. Eur., 99.7-100.5% (oxidimetric)
Sigma-Aldrich
L-Ascorbic acid, BioUltra, ≥99.5% (RT)
Sigma-Aldrich
L-Ascorbic acid, powder, suitable for cell culture, γ-irradiated
Sigma-Aldrich
L-Ascorbic acid, reagent grade
Sigma-Aldrich
L-Ascorbic acid, reagent grade, crystalline
Sigma-Aldrich
L-Ascorbic acid, BioXtra, ≥99.0%, crystalline
Sigma-Aldrich
L-Ascorbic acid, suitable for cell culture, suitable for plant cell culture, ≥98%
Sigma-Aldrich
3-Isobutyl-1-methylxanthine, BioUltra, ≥99%
Sigma-Aldrich
3-Isobutyl-1-methylxanthine, ≥99% (HPLC), powder
Sigma-Aldrich
L-Ascorbic acid, meets USP testing specifications
Sigma-Aldrich
MISSION® esiRNA, targeting mouse Ntrk3
Sigma-Aldrich
MISSION® esiRNA, targeting human NTRK3 (2)
Sigma-Aldrich
Fluorescein, suitable for fluorescence, free acid
Sigma-Aldrich
Dexamethasone, powder, BioReagent, suitable for cell culture, ≥97%
Sigma-Aldrich
Dexamethasone, powder, γ-irradiated, BioXtra, suitable for cell culture, ≥80% (HPLC)
Sigma-Aldrich
Dexamethasone, meets USP testing specifications
Sigma-Aldrich
Dexamethasone, ≥98% (HPLC), powder
Sigma-Aldrich
DAPI, for nucleic acid staining